Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

SAB4200322

Sigma-Aldrich

Anti-OSTF1 (N-terminal) antibody produced in rabbit

~1.0 mg/mL, affinity isolated antibody

Synonym(s):

Anti-OSF1, Anti-Osteoclast-stimulating factor 1

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

rabbit

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

antigen ~26 kDa

species reactivity

human

concentration

~1.0 mg/mL

technique(s)

immunoprecipitation (IP): 2.5-5.0 μg using lysates of HEK-293T cells over expressing human OSTF1
western blot: 2-4 μg/mL using using HEK-293 cells overexpressing human OSTF1

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... OSTF1(26578)
mouse ... Ostf1(20409)
rat ... Ostf1(259275)

General description

Osteoclast-stimulating factor (OSF or OSTF1) is a small intracellular protein with a N-terminal proline-rich region, SRC Homology 3 (SH3) domain, three ankyrin repeats and a C-terminal aspartate-rich region. The OSF gene is mapped to human chromosome 9q21.13. OSF is expressed in osteoclast (OCL) as well as motor neurons. Structurally, OCF protein takes up a β-barrel fold.

Specificity

Anti-OSTF1 (N-terminal) recognizes human OSTF1.

Immunogen

synthetic peptide corresponding to the N-terminal region of human OSTF1, conjugated to KLH. The corresponding sequence is identical in rat and mouse.

Application

Anti-OSTF1 (N-terminal) antibody produced in rabbit may be used in immunoblotting and immunoprecipitation.

Biochem/physiol Actions

Osteoclast-stimulating factor (OSF or OSTF1) SRC Homology 3 (SH3) domain undergoes conformational changes to bind to proline-rich ligand. The C-terminal ankyrin repeats preferably binds to negative-charged binding partners. OSF plays an indirect role in osteoclast (OCL) formation as well as bone resorption. It favors OCL formation by interacting with sarcoma proto oncogene (c-Src) and its associated protein-based signaling pathway. OSF is implicated in the pathophysiology of spinal muscular atrophy by its possible interaction with survival motor neuron (SMN) proteins through the SH3 domain. OSF-SH3 domain interaction stimulates OCL formation and its inhibition may reduce bone resorption by OCL. A compromised bone resorption functionality in OCL further triggers bone loss and may lead to various bone disorders, including hyperparathyroidism, Paget′s disease, osteoporosis, estrogen deficiency, periodontal disease as well as inflammatory bone loss and bone malignancy.

Physical form

Solution in 0.01 M phos­phate buffered saline, pH 7.4, containing 15 mM sodium azide.

Storage and Stability

For continuous use, store at 2-8 °C for up to one month. For extended storage, freeze in working aliquots. Repeated freezing and thawing, or storage in “frost-free” freezers,is not recommended. If slight turbidity occurs upon prolonged storage, clarify the solution by centrifugation before use. Working dilutions should be discarded if not used within 12 hours.

Disclaimer

Unless otherwise stated in our catalog, our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class

10 - Combustible liquids

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Structure of the SH3 domain of human osteoclast-stimulating factor at atomic resolution
Chen L, et al.
Acta Crystallographica Section F, Structural Biology and Crystallization Communications, 844-844 (2006)
Matthieu Vermeren et al.
Mammalian genome : official journal of the International Mammalian Genome Society, 28(11-12), 498-514 (2017-09-25)
Osteoclast stimulation factor 1 (OSTF1) is an SH3-domain containing protein that was initially identified as a factor involved in the indirect activation of osteoclasts. It has been linked to spinal muscular atrophy in humans through its interaction with SMN1, and
Crystal structure of human osteoclast stimulating factor.
Shuilong Tong et al.
Proteins, 75(1), 245-251 (2009-01-13)
N Kurihara et al.
The Journal of biological chemistry, 276(44), 41035-41039 (2001-09-12)
We have recently identified and cloned an intracellular peptide termed osteoclast-stimulating factor (OSF) that increases osteoclast (OCL) formation and bone resorption through a cellular signal transduction cascade, possibly through its interaction with c-Src or related family members. To further identify

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service