Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

SAB4200317

Sigma-Aldrich

Anti-OS9 antibody, Mouse monoclonal

clone OS9-8, purified from hybridoma cell culture

Synonym(s):

Anti-ERLEC2, Anti-Endoplasmic reticulum lectin 2, Anti-Endoplasmic reticulum lectin, ERLEC2, Anti-Erlectin 2, Anti-OS-9, amplified in osteosarcoma 9, Anti-Osteosarcoma amplified 9

Sign Into View Organizational & Contract Pricing

Select a Size

200 μL
$401.00

$401.00


Check Cart for Availability

Request a Bulk Order
Notify Me

Get notified when this item is ready to ship via email.


Select a Size

Change View
200 μL
$401.00

About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

$401.00


Check Cart for Availability

Request a Bulk Order
Notify Me

Get notified when this item is ready to ship via email.

biological source

mouse

conjugate

unconjugated

antibody form

purified from hybridoma cell culture

antibody product type

primary antibodies

clone

OS9-8, monoclonal

form

buffered aqueous solution

mol wt

antigen ~97/88 kDa

species reactivity

human, rat, mouse

concentration

~1.0 mg/mL

technique(s)

immunoprecipitation (IP): suitable
western blot: 2-4 μg/mL using HEK-293T cells overexpressing OS9.

isotype

IgG2b

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... OS9(10956)
mouse ... Os9(216440)
rat ... Os9(362891)

General description

Monoclonal Anti-OS9 (mouse IgG2b isotype) is derived from the hybridoma OS9-8 produced by the fusion of mouse myeloma cells and splenocytes from BALB/c mice immunized with a synthetic peptide. Amplified in osteosarcoma 9 (OS-9) is a N-glycosylated protein, mapped to human chromosome 12q13.3-q14.1. It is ubiquitously expressed in two isoforms OS-9.1 and OS-9.2.

Specificity

Monoclonal Anti-OS9 recognizes human, rat and mouse OS9 isoforms 1 and 2.

Immunogen

synthetic peptide corresponding to the C-terminal region of human OS9, conjugated to KLH. The corresponding sequence is identical in mouse and rat OS9. The antibody recognizes OS9 isoforms 1 and 2.

Application

Monoclonal Anti-OS9 antibody produced in mouse may be used in immunoblotting and immunoprecipitation.

Biochem/physiol Actions

Amplified in osteosarcoma 9 (OS-9) is an endoplasmic reticulum (ER)-resident lectin involved in ER quality control. OS9 is upregulated in response to ER stress through activation of the inositol-requiring enzyme 1/X-box binding protein 1 (Ire1/Xbp1) pathway. OS9 associates both with components of the Endoplasmic-reticulum-associated protein degradation (ERAD) machinery and ERAD substrates. It binds to misfolding proteins preventing their secretion from the ER and directing them to the suppressor/enhancer of Lin-12-like (SEL1L)/HMG-CoA reductase degradation protein 1 (HRD1) dislocation and ubiquitylation complex in the ER membrane. Two OS9 spliced variants are amplified in tumors.

Physical form

Solution in 0.01 M phosphate buffered saline, pH 7.4, containing 15 mM sodium azide.

Storage and Stability

For continuous use, store at 2-8°C for up to one month. For extended storage, freeze at -20oC in working aliquots. Repeated freezing and thawing,or storage in “frost-free” freezers,is not recommended. If slight turbidity occurs upon prolonged storage, clarify the solution by centrifugation before use. Working dilution samples should be discarded if not used within 12 hours.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class

10 - Combustible liquids

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Chromosomal and genetic imbalances in Chinese patients with rhabdomyosarcoma detected by high-resolution array comparative genomic hybridization
Liu C, et al.
International Journal of Clinical and Experimental Pathology, 7(2), 690-690 (2014)
Stringent requirement for HRD1, SEL1L, and OS-9/XTP3-B for disposal of ERAD-LS substrates
Bernasconi R, et al.
The Journal of cell biology, 188(2), 223-235 (2010)
A Dual Task for the Xbp1-responsive OS-9 Variants in the Mammalian Endoplasmic Reticulum INHIBITING SECRETION OF MISFOLDED PROTEIN CONFORMERS AND ENHANCING THEIR DISPOSAL
Bernasconi R, et al.
The Journal of biological chemistry, 283(24), 16446-16454 (2008)
Nobuko Hosokawa et al.
The Journal of biological chemistry, 284(25), 17061-17068 (2009-04-07)
In the endoplasmic reticulum (ER), lectins and processing enzymes are involved in quality control of newly synthesized proteins for productive folding as well as in the ER-associated degradation (ERAD) of misfolded proteins. ER quality control requires the recognition and modification

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service