Skip to Content
MilliporeSigma
All Photos(3)

Key Documents

SAB4200315

Sigma-Aldrich

Anti-EAPP (E2F-Associated Phosphoprotein) antibody, Mouse monoclonal

clone 1E4, purified from hybridoma cell culture

Synonym(s):

Anti-BM036

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

mouse

conjugate

unconjugated

antibody form

purified from hybridoma cell culture

antibody product type

primary antibodies

clone

1E4, monoclonal

form

buffered aqueous solution

mol wt

antigen ~42 kDa

species reactivity

human, mouse

concentration

~1.0 mg/mL

technique(s)

indirect immunofluorescence: suitable
western blot: 2.5-5.0 μg/mL using whole extracts of human HEK-293T cells

isotype

IgG1

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... EAPP(55837)

General description

Monoclonal Anti-EAPP (E2F-associated phosphoprotein) (mouse IgG1 isotype) is derived from the hybridoma 1E4 produced by the fusion of mouse myeloma cells and splenocytes from BALB/c mice immunized with a fusion protein. The EEAP gene is mapped to human chromosome 14q13.1. The encoded protein localizes to the nucleus and is present throughout the cell cycle except during mitosis.

Specificity

Monoclonal Anti-EAPP (E2F-associated phosphoprotein) recognizes human and mouse EAPP.

Immunogen

fusion protein corresponding to the N-terminus of human EAPP

Application

Anti-EAPP (E2F-Associated Phosphoprotein) antibody, Mouse monoclonal is suitable for immunoblotting and immunofluorescence.

Biochem/physiol Actions

E2F-associated phosphoprotein (EAPP) interacts with several members of the E2F family of proteins. The E2F family plays a crucial role in the control of the cell cycle and in the action of tumor suppressor proteins. It functions to modulate E2F-regulated transcription and stimulates proliferation. Increased levels of EEAP is observed in most transformed human cells.

Physical form

Solution in 0.01 M phosphate buffered saline, pH 7.4, containing 15 mM sodium azide.

Storage and Stability

For continuous use, store at 2-8°C for up to one month. For extended storage, freeze at -20oC in working aliquots. Repeated freezing and thawing,or storage in “frost-free” freezers,is not recommended. If slight turbidity occurs upon prolonged storage, clarify the solution by centrifugation before use. Working dilution samples should be discarded if not used within 12 hours.

Disclaimer

Unless otherwise stated in our catalog, our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class

10 - Combustible liquids

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Yang Xue et al.
Journal of cranio-maxillo-facial surgery : official publication of the European Association for Cranio-Maxillo-Facial Surgery, 43(7), 1248-1255 (2015-07-01)
To report on the molecular genetic analysis of a Chinese patient with Maffucci syndrome. Using the genomic DNA extracted from the patient's hemangioma sample, the coding exons and exon/intron splice junctions of the IDH1 and IDH2 genes were amplified by
Michael Novy et al.
Molecular biology of the cell, 16(5), 2181-2190 (2005-02-18)
E2F transcription factors play an essential role in cell proliferation and apoptosis and their activity is frequently deregulated in human cancers. In a yeast two-hybrid screen we identified a novel E2F-binding protein. Due to its strong phosphorylation we named it
Ludwig Schwarzmayr et al.
The international journal of biochemistry & cell biology, 40(12), 2845-2853 (2008-07-01)
The E2F-associated phosphoprotein (EAPP) is a ubiquitous nuclear protein that interacts with the activating members of the E2F family of transcription factors and increases the activity of several cell-cycle regulated promoters in an E2F-dependent manner. Our previous studies also showed

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service