Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

SAB4200314

Sigma-Aldrich

Anti-PRR5 (C-terminal) antibody produced in rabbit

~1.0 mg/mL, affinity isolated antibody

Synonym(s):

Anti-PROTOR-1, Anti-Proline rich 5 (renal), PP610, Anti-Proline-rich 5

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

rabbit

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

antigen ~45/50 kDa

species reactivity

human

concentration

~1.0 mg/mL

technique(s)

indirect immunofluorescence: 10-20 μg/mL using human HeLa cells.
western blot: 3-6 μg/mL using whole extracts of human HEK-293T or HepG2 cells.

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... PRR5(55615)

General description

The proline-rich protein 5 (PRR5) gene is mapped to human chromosome 22q13.31. PRR5, also known as Protor-1, is a conserved proline-rich protein highly expressed in kidney. PRR5 is a component of the mammalian target of rapamycin complex 2 (mTORC2). Alternative splicing of the gene gives rise to five transcripts encoding different isoforms.

Specificity

Anti-PRR5 recognizes human PRR5.

Immunogen

Solution in 0.01 M phosphate buffered saline pH 7.4, containing 15 mM sodium azide.

Application

Anti-PRR5 (C-terminal) antibody produced in rabbit is suitable for immunoblotting.

Biochem/physiol Actions

The proline-rich protein 5 (PRR5) gene acts as a tumor suppressor gene that may be involved in breast and colorectal tumorigenesis. It regulates platelet-derived growth factor (PDGF) receptor β expression and PDGF signaling to protein kinase B (Akt) and S6 Kinase 1 (S6K1).

Physical form

synthetic peptide corresponding to the C-terminal region of human PRR5, conjugated to KLH. The corresponding sequence differs by a single amino acid in mouse and by 2 amino acids in rat.

Storage and Stability

For continuous use, store at 2-8 °C for up to one month. For extended storage freeze in working aliquots. Repeated freezing and thawing is not recommended.If slight turbidity occurs upon prolonged storage, clarify the solution by centrifugation before use. Working dilution samples should be discarded if not used within 12 hours.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class

10 - Combustible liquids

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

mTORC2: The other facet of mTOR
Treins C and Downward J
The Enzymes null
Cameron N Johnstone et al.
Genomics, 85(3), 338-351 (2005-02-19)
Loss of heterozygosity on chromosome 22q13.31 is a frequent event during human breast and colorectal carcinogenesis. Herein we characterize a novel gene at chromosome 22q13.31 designated PRR5. Alternative promoter usage and splicing converge to generate five PRR5 transcript variants with
So-Yon Woo et al.
The Journal of biological chemistry, 282(35), 25604-25612 (2007-06-30)
The protein kinase mammalian target of rapamycin (mTOR) plays an important role in the coordinate regulation of cellular responses to nutritional and growth factor conditions. mTOR achieves these roles through interacting with raptor and rictor to form two distinct protein

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service