Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

SAB4200313

Sigma-Aldrich

Anti-PRR5 antibody produced in rabbit

~1.0 mg/mL, affinity isolated antibody

Synonym(s):

Anti-PROTOR-1, Anti-PROTOR1, Anti-Proline rich 5 (renal), PP610

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

rabbit

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

antigen ~45/50 kDa

species reactivity

human

concentration

~1.0 mg/mL

technique(s)

western blot: 0.5-1.0 μg/mL using whole extracts of HEK-293T cells over expressing human PRR5.

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... PRR5(55615)
mouse ... Prr5(109270)
rat ... Prr5(315189)

General description

Proline-rich protein 5 (PRR5), also known as Protor-1, is a proline-rich protein expressed abundantly in kidney and is conserved. PRR5 is a component of the mammalian target of rapamycin complex 2 (mTORC2). Alternative splicing and the use of alternative promoters results in five transcripts encoding different isoforms. The PRR5 gene is mapped to human chromosome 22q13.31.

Specificity

Anti-PRR5 recognizes human PRR5.

Immunogen

synthetic peptide corresponding to an internal region of human PRR5, conjugated to KLH. The corresponding sequence is identical in mouse and rat

Application

Anti-PRR5 antibody produced in rabbit is suitable for immunoblotting.

Biochem/physiol Actions

The proline-rich protein 5 (PRR5) gene exhibits tumor suppression action. It may be associated with breast and colorectal tumorigenesis. PRR5 controls the expression of platelet-derived growth factor (PDGF) receptor β and mediates PDGF signaling to protein kinase B (Akt) and S6 Kinase 1 (S6K1).

Physical form

Solution in 0.01 M phosphate buffered saline pH 7.4, containing 15 mM sodium azide.

Storage and Stability

For continuous use, store at 2-8 °C for up to one month. For extended storage freeze in working aliquots. Repeated freezing and thawing is not recommended. If slight turbidity occurs upon prolonged storage, clarify the solution by centrifugation before use. Working dilution samples should be discarded if not used within 12 hours.

Disclaimer

Unless otherwise stated in our catalog, our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class

10 - Combustible liquids

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

mTORC2: The other facet of mTOR
The Enzymes null
So-Yon Woo et al.
The Journal of biological chemistry, 282(35), 25604-25612 (2007-06-30)
The protein kinase mammalian target of rapamycin (mTOR) plays an important role in the coordinate regulation of cellular responses to nutritional and growth factor conditions. mTOR achieves these roles through interacting with raptor and rictor to form two distinct protein
Cameron N Johnstone et al.
Genomics, 85(3), 338-351 (2005-02-19)
Loss of heterozygosity on chromosome 22q13.31 is a frequent event during human breast and colorectal carcinogenesis. Herein we characterize a novel gene at chromosome 22q13.31 designated PRR5. Alternative promoter usage and splicing converge to generate five PRR5 transcript variants with

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service