Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

SAB4200308

Sigma-Aldrich

Anti-Synaptotagmin 7 (C-terminal) antibody produced in rabbit

enhanced validation

~1.5 mg/mL, affinity isolated antibody

Synonym(s):

Anti-IPCA-7, Anti-PCANAP7, Anti-SYT-VII, Anti-SYT7

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

rabbit

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

antigen ~45 kDa

species reactivity

human, rat, mouse

enhanced validation

recombinant expression
Learn more about Antibody Enhanced Validation

concentration

~1.5 mg/mL

technique(s)

western blot: 2-4 μg/mL using mouse brain extracts (S1 fraction), rat brain extracts (S1 fraction) or HEK-293T cell lysates over expressing human synaptotagmin 7

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... SYT7(9066)
mouse ... Syt7(54525)
rat ... Syt7(59267)

General description

Synaptotagmin7 (SYT7) belongs to the synaptotagmin gene family. It codes for and encodes a single-pass 46-kDa transmembrane protein and is located on human chromosome11q12.2. Syt7 contains a single transmembrane domain, a variable linker sequence and C2A and C2B Ca2+-binding domains. It is ubiquitously expressed during development, but is later restricted to dividing cells, postmitotic neurons and neuroendocrine cells.

Immunogen

synthetic peptide corresponding to a sequence located near the C-terminus of human synaptotagmin 7 (Syt7), conjugated to KLH. The corresponding sequence is identical in rat and mouse Syt7.

Application

Anti-Synaptotagmin 7 (C-terminal) antibody produced in rabbit has been used in immunoblotting (~45 kDa).

Biochem/physiol Actions

Synaptotagmin7 (SYT7) helps to facilitate the calciumdependent modulation of membrane trafficking during synaptic transmission. It has been proposed to regulate different types of Ca2+ -dependent exocytosis. Syt7 has been proposed to play a key role in the regulation of Ca2+- dependent lysosome exocytosis. Knockout of Syt7 in mice results in impaired insulin secretion and glucose intolerance.

Physical form

Solution in 0.01 M phos­phate buffered saline, pH 7.4, containing 15 mM sodium azide.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class

10 - Combustible liquids

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Downregulation of SYT7 inhibits glioblastoma growth by promoting cellular apoptosis
Xiao B, et al.
Molecular Medicine Reports, 16(6), 9017-9022 (2017)
SYT7 acts as a driver of hepatic metastasis formation of gastric cancer cells
Kanda M, et al.
Oncogene, 37(39), 5355-5355 (2018)
Genetic analysis of synaptotagmin-7 function in synaptic vesicle exocytosis
Maximov A, et al.
Proceedings of the National Academy of Sciences of the USA, 105(10), 3986-3991 (2008)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service