Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

SAB4200305

Sigma-Aldrich

Anti-mLST8/GBL antibody produced in rabbit

~1.0 mg/mL, affinity isolated antibody

Synonym(s):

Anti-G protein beta subunit-like, Anti-GBL, Anti-Gable, Anti-GbetaL, Anti-LST8, Anti-MGC111011, Anti-MLST8, Anti-MTOR associated protein, Anti-POP3, Anti-TORC subunit LST8, Anti-Target of rapamycin complex subunit LST8, Anti-WAT1, Anti-mammalian lethal with SEC13 protein 8

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

rabbit

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

antigen ~38 kDa

species reactivity

human

concentration

~1.0 mg/mL

technique(s)

western blot: 2-4 μg/mL using whole extracts of HEK-293T cells over expressing human mLST8.

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... MLST8(64223)
mouse ... Mlst8(56716)
rat ... Mlst8(64226)

General description

Mammalian lethal with SEC13 protein 8 (mLST8), also known as GβL (G protein β-subunit-like protein), is a widely expressed protein. It consists of seven WD40 repeats (tryptophan-aspartic acid/ beta-transducing repeats) and shares great sequence homology with the β subunits of heterotrimeric G proteins. mLST8 is conserved among all eukaryotes. mLST8 is a subunit of the mammalian target of rapamycin (mTOR) signaling complexes, mTORC1 and mTORC2. The MLST8 gene is mapped to human chromosome 16p13.3.

Specificity

Anti-mLST8/GBL recognizes human mLST8.

Immunogen

synthetic peptide corresponding to an internal region within human mLST8, conjugated to KLH. The corresponding sequence is identical in mouse, rat, canine, bovine and monkey.

Application

Anti-mLST8/GBL antibody produced in rabbit is suitable for immunoblotting.

Biochem/physiol Actions

Mammalian lethal with SEC13 protein 8 (mLST8) associates with the kinase domain of mammalian target of rapamycin (mTOR) and stimulates mTOR kinase activity. mLST8 is significantly involved in the nutrient sensitive raptor-mTOR interaction. It is also implicated in the raptor-mediated inhibition of mTOR kinase activity. mLST8 expression is essential for the phosphorylation of S6 Kinase 1 (S6K1), in vivo. This infers that mLST8 positively regulates the mTOR pathway. Upregulation of mLST8 has been observed in hepatocellular carcinoma (HCC).

Physical form

Solution in 0.01 M phosphate buffered saline pH 7.4, containing 15 mM sodium azide.

Storage and Stability

For continuous use, store at 2-8°C for up to one month. For extended storage freeze in working aliquots. Repeated freezing and thawing is not recommended. If slight turbidity occurs upon prolonged storage, clarify the solution by centrifugation before use. Working dilution samples should be discarded if not used within 12 hours.

Disclaimer

Unless otherwise stated in our catalog, our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class

10 - Combustible liquids

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Do-Hyung Kim et al.
Molecular cell, 11(4), 895-904 (2003-04-30)
mTOR and raptor are components of a signaling pathway that regulates mammalian cell growth in response to nutrients and growth factors. Here, we identify a member of this pathway, a protein named GbetaL that binds to the kinase domain of
Xiang-Nan Yu et al.
Annals of surgical oncology, 27(5), 1546-1557 (2020-03-12)
The mechanistic target of rapamycin (mTOR) pathway, containing mTOR complex 1 (mTORC1) and mTORC2, is dysregulated in multiple cancers, including hepatocellular carcinoma (HCC). Mammalian lethal with sec-13 protein 8 (mLST8) is a shared constituent of both mTORC1 and mTORC2, yet
David A Guertin et al.
Developmental cell, 11(6), 859-871 (2006-12-05)
The mTOR kinase controls cell growth, proliferation, and survival through two distinct multiprotein complexes, mTORC1 and mTORC2. mTOR and mLST8 are in both complexes, while raptor and rictor are part of only mTORC1 and mTORC2, respectively. To investigate mTORC1 and
Ake T Lu et al.
Nature communications, 7, 10561-10561 (2016-02-03)
DNA methylation (DNAm) levels lend themselves for defining an epigenetic biomarker of aging known as the 'epigenetic clock'. Our genome-wide association study (GWAS) of cerebellar epigenetic age acceleration identifies five significant (P<5.0 × 10(-8)) SNPs in two loci: 2p22.1 (inside

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service