Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

SAB4200303

Sigma-Aldrich

Anti-YY1 (N-terminal) antibody produced in rabbit

Ig fraction of antiserum

Synonym(s):

Anti-Transcriptional repressor protein YY1, Delta transcription factor, NF-E1, INO80S, Anti-Yin and Yang 1

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

rabbit

conjugate

unconjugated

antibody form

Ig fraction of antiserum

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

antigen ~55 kDa

species reactivity

human

technique(s)

western blot: 1:2000-1:4000 using HEK-293T cells over-expressing human YY1

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... YY1(7528)
rat ... Yy1(24919)

General description

The Yin Yang-1 (YY1) gene is mapped to human chromosome 14q32.2 and encodes a ubiquitous and conserved transcription factor. It is a Gli-Kruppel type zinc finger protein containing four C2H2-type zinc-finger motifs and two specific domains as an activator or a repressor. It belongs to the polycomb group protein family. Alternative splicing of this gene yields eight different transcripts.

Specificity

Anti-YY1 (N-terminal) recognizes human YY1.

Immunogen

synthetic peptide corresponding to the N- terminal region of human YY1, conjugated to KLH. The corresponding sequence is identical in rat.

Application

Anti-YY1 (N-terminal) antibody produced in rabbit has been used in immunoblotting.[1]

Biochem/physiol Actions

The N-terminus region of Yin Yang-1 (YY1) protein controls the activation, repression and protein-protein interaction functions. The YY1 protein is shown to regulate several genes including viral and cellular genes. It is essential for cell growth, development, differentiation and tumor suppression. The YY1 gene is found to be upregulated in association with cellular proliferation, resistance to Fas ligand-mediated apoptosis, tumorigenesis and metastatic prostate cancer. YY1 is closely associated with hematopoiesis and cell cycle control.

Physical form

Solution in 0.01 M phos­phate buffered saline, pH 7.4, containing 15 mM sodium azide.

Storage and Stability

For continuous use, store at 2-8 °C for up to one month. For extended storage, freeze in working aliquots. Repeated freezing and thawing, or storage in “frost-free” freezers,is not recommended. If slight turbidity occurs upon prolonged storage, clarify the solution by centrifugation before use. Working dilutions should be discarded if not used within 12 hours.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class

10 - Combustible liquids

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Chunqing Luo et al.
Genomics, 87(3), 348-355 (2005-12-27)
YY2 was originally identified due to its unusual similarity to the evolutionarily well-conserved zinc finger gene YY1. In this study, we have determined the evolutionary origin and conservation of YY2 using comparative genomic approaches. Our results indicate that YY2 is
Giancarlo Castellano et al.
Cell cycle (Georgetown, Tex.), 8(9), 1367-1372 (2009-04-04)
The Yin Yang 1 (YY1) transcription factor has a pivotal role in normal biological processes such as development, differentiation, replication and cell proliferation exerting its effects on a huge number of genes involved in these processes. Mechanisms of YY1 action
Soroush Tahmasebi et al.
Proceedings of the National Academy of Sciences of the United States of America, 113(44), 12360-12367 (2016-11-03)
Translational control of gene expression plays a key role during the early phases of embryonic development. Here we describe a transcriptional regulator of mouse embryonic stem cells (mESCs), Yin-yang 2 (YY2), that is controlled by the translation inhibitors, Eukaryotic initiation

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service