Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

SAB4200302

Sigma-Aldrich

Anti-YY1 antibody produced in rabbit

IgG fraction of antiserum

Synonym(s):

Anti-Delta transcription factor, Anti-INO80S, Anti-NF-E1, Anti-Transcriptional repressor protein YY1, Anti-Yin and Yang 1

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

rabbit

conjugate

unconjugated

antibody form

IgG fraction of antiserum

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

antigen ~55 kDa

species reactivity

rat, human

technique(s)

western blot: 1:1000-1:2000 using lysates of HEK-293T cells over expressing human YY1.

UniProt accession no.

shipped in

dry ice

Storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... YY1(7528)
rat ... Yy1(24919)

General description

The Yin Yang-1 (YY1) gene encodes a ubiquitous, conserved, multifunctional zinc-finger transcription factor. It is a phosphoprotein and a member of the polycomb group protein family. The YY1 gene is mapped to human chromosome 14q32.2. Alternative splicing of the gene produces eight different transcripts. The YY1 protein possesses four C2H2-type zinc-finger motifs and two specific domains as an activator or a repressor.

Specificity

Anti-YY1 recognizes human YY1.

Immunogen

synthetic peptide corresponding to an Internal region of human YY1, conjugated to KLH. The corresponding sequence is identical in rat.

Application

Anti-YY1 antibody produced in rabbit is has been used in immunofluorescence[1]. It is suitable for immunoblotting.

Biochem/physiol Actions

Yin Yang-1 gene (YY1) significantly participates in hematopoiesis and cell cycle control. The YY1 protein regulates initiation, activation, or repression of transcription of a variety of genes required for cell growth, development, differentiation, or tumor suppression, depending upon the context in which it binds. YY1 overexpression is associated with cellular proliferation, resistance to Fas ligand-mediated apoptosis, tumorigenesis and metastatic prostate cancer.

Physical form

Solution in 0.01 M phos­phate buffered saline, pH 7.4, containing 15 mM sodium azide.

Storage and Stability

For continuous use, store at 2-8 °C for up to one month. For extended storage, freeze in working aliquots. Repeated freezing and thawing, or storage in “frost-free” freezers,is not recommended. If slight turbidity occurs upon prolonged storage, clarify the solution by centrifugation before use. Working dilutions should be discarded if not used within 12 hours.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class

10 - Combustible liquids

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Giancarlo Castellano et al.
Cell cycle (Georgetown, Tex.), 8(9), 1367-1372 (2009-04-04)
The Yin Yang 1 (YY1) transcription factor has a pivotal role in normal biological processes such as development, differentiation, replication and cell proliferation exerting its effects on a huge number of genes involved in these processes. Mechanisms of YY1 action
Isha Sharma et al.
Biomedicines, 8(7) (2020-07-28)
Obesity is associated with perturbations in cellular energy homeostasis and consequential renal injury leading to chronic renal disease (CKD). Myo-inositol oxygenase (MIOX), a tubular enzyme, alters redox balance and subsequent tubular injury in the settings of obesity. Mechanism(s) for such

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service