Skip to Content
MilliporeSigma
All Photos(3)

Key Documents

SAB4200289

Sigma-Aldrich

Anti-Dystrobrevin-β (N-terminal) antibody produced in rabbit

~1.5 mg/mL, affinity isolated antibody

Synonym(s):

Anti-DTNB

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

rabbit

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

antigen ~72 kDa

species reactivity

rat, human

concentration

~1.5 mg/mL

technique(s)

immunohistochemistry: 20-40 μg/mL using formalin-fixed, paraffin-embedded rat kidney.
indirect immunofluorescence: 2-4 μg/mL using HEK-293T cells overexpressing human dystrobrevin-β.
western blot: 1.5-3.0 μg/mL using lysates of HEK-293T cells overexpressing human dystrobrevin-β.

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... DTNB(1838)
rat ... Dtnb(362715)

General description

The dystrobrevin-β(DTNB) gene is mapped to human chromosome 2p23.3. The encoded protein belongs to the dystrobrevin (DB) subfamily of the dystrophin-related protein family. DB is a component of the dystrophin-glycoprotein complex (DGC) that is located at the muscle sarcolemma and forms a transmembrane link between the cytoskeleton and the basal lamina. β-DB is confined to non-muscle tissues and predominantly expressed in the kidney and brain. β-DB is linked to dystrophin and syntrophin isoforms in the hippocampus, cortex, and Purkinje cells. It is abundant in the post-synaptic densities (PSDs).

Specificity

Anti-Dystrobrevin-b(N-terminal) specifically recognizes human and rat dystrobrevin-b.

Immunogen

synthetic peptide corresponding to a sequence located at the N-terminus of human dystrobrevin-b (DTNB), conjugated to KLH. The corresponding sequence is identical in human DTNB isoforms 1-5, and highly conserved (single amino acid substitution) in rat and mouse DTNB.

Application

Anti-Dystrobrevin-β (N-terminal) antibody produced in rabbit is suitable for:
  • immunoblotting
  • immunofluorescence
  • immunohistochemistry

Biochem/physiol Actions

Dystrobrevin-β (β-DB) may be significantly involved in assembling and supporting the dystrophin-associated protein complex (DPC)-like complexes in non-muscle tissues. It is known to interact with the neuronal kinesin heavy chain Kif5A and Kif5B in the brain, suggesting that β-DB might act as a motor protein receptor involved in the transport of dystrophin-glycoprotein complex (DGC) components to specific sites in the cell.

Physical form

Solution in 0.01 M phos­phate buffered saline, pH 7.4, containing 15 mM sodium azide.

Storage and Stability

For continuous use, store at 2-8 °C for up to one month. For extended storage, freeze in working aliquots. Repeated freezing and thawing, or storage in “frost-free” freezers,is not recommended. If slight turbidity occurs upon prolonged storage, clarify the solution by centrifugation before use. Working dilutions should be discarded if not used within 12 hours.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class

10 - Combustible liquids

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Maria Santa Rocca et al.
European journal of medical genetics, 56(1), 62-65 (2012-11-13)
Deletions of the short arm of chromosome 2 are exceedingly rare and only nine cases involving regions from 2p23 to 2pter have been reported to date. Most of these deletions had only been analysed by GTG banding. Here, we report
P Macioce et al.
Journal of cell science, 116(Pt 23), 4847-4856 (2003-11-06)
Beta-dystrobrevin, a member of the dystrobrevin protein family, is a dystrophin-related and -associated protein restricted to non-muscle tissues and is highly expressed in kidney, liver and brain. Dystrobrevins are now thought to play an important role in intracellular signal transduction
T Haenggi et al.
Cellular and molecular life sciences : CMLS, 63(14), 1614-1631 (2006-05-20)
The dystrophin glycoprotein complex (DGC) is a multimeric protein assembly associated with either the X-linked cytoskeletal protein dystrophin or its autosomal homologue utrophin. In striated muscle cells, the DGC links the extracellular matrix to the actin cytoskeleton and mediates three

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service