Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

SAB4200286

Sigma-Aldrich

Anti-LYN antibody, Mouse monoclonal

clone LYN-27, purified from hybridoma cell culture

Synonym(s):

Monoclonal Anti-JTK8, Monoclonal Anti-v-yes-1 Yamaguchi sarcoma viral related oncogene homolog

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

mouse

conjugate

unconjugated

antibody form

purified from hybridoma cell culture

antibody product type

primary antibodies

clone

LYN-27, monoclonal

form

buffered aqueous solution

mol wt

antigen ~58 kDa

species reactivity

human, mouse

concentration

~1.0 mg/mL

technique(s)

immunocytochemistry: suitable
western blot: 2.0-4.0 μg/mL using extracts of HL-60 cells

isotype

IgG2b

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... LYN(4067)
mouse ... Lyn(17096)

General description

Monoclonal Anti-LYN (mouse IgG2b isotype) is derived from the hybridoma LYN-27 produced by the fusion of mouse myeloma cells and splenocytes from BALB/c mice immunized with a peptide. The LYN gene is mapped to human chromosome 8q12.1. Lyn is an important Src-family kinase (SFK) expressed in B lymphocytes. Lyn contains an N-terminal unique domain, SH3, SH2, and tyrosine kinases domain. Lyn exists as two isoforms, p53 and p56, arising from alternate splicing of exon 2.

Specificity

Monoclonal Anti-LYN recognizes human and mouse LYN.

Immunogen

peptide corresponding to a sequence within human LYN, conjugated to KLH. This sequence is identical in human LYN A and B isoforms, and highly conserved in rat and mouse LYN (94%)

Application

Anti-LYN antibody, Mouse monoclonal is suitable for immunoblotting and immunocytochemistry.

Biochem/physiol Actions

The tyrosine-protein kinase Lyn acts as both a positive and negative regulator of B cell receptor signaling pathways. Interestingly, it is a critical mediator in many pathways of B cell activation through the cluster of differentiation 19 (CD19) and CD180. It is shown that Lyn activity is associated with the pathological conditions arising due to the LYN gene related loss of function and gain of function in mice. It might cause lethal antibody-mediated autoimmune diseases and myeloid neoplasia in mice.

Physical form

Solution in 0.01 M phosphate buffered saline, pH 7.4, containing 15 mM sodium azide.

Storage and Stability

For continuous use, store at 2-8°C for up to one month. For extended storage, freeze at -20oC in working aliquots. Repeated freezing and thawing,or storage in “frost-free” freezers,is not recommended. If slight turbidity occurs upon prolonged storage, clarify the solution by centrifugation before use. Working dilution samples should be discarded if not used within 12 hours.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class

10 - Combustible liquids

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Yuekang Xu et al.
Immunity, 22(1), 9-18 (2005-01-25)
Lyn, one of several Src-family tyrosine kinases in immune cells, is noted for its ability to negatively regulate signaling pathways through phosphorylation of inhibitory receptors, enzymes, and adaptors. Somewhat paradoxically, it is also a key mediator in several pathways of
Clifford A Lowell
Molecular immunology, 41(6-7), 631-643 (2004-06-29)
In immune cells, Src-family kinases have been implicated as critical regulators of a large number of intracellular signaling pathways. Studies of gene knockout mice lacking various family members have both confirmed the requirement for these kinases in some pathways and
Edmond S K Ma et al.
Cancer genetics, 218-219, 15-19 (2017-11-21)
In a newly diagnosed patient with acute myeloid leukemia (AML) and complex cytogenetics and negative for gene mutations associated with myeloid neoplasms, RNA sequencing by next-generation sequencing (NGS) through a large cancer-related gene panel showed ETV6-LYN leukemic fusion transcript. Breakpoint

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service