Skip to Content
MilliporeSigma
All Photos(3)

Key Documents

SAB4200285

Sigma-Aldrich

Anti-VHL (C-terminal) antibody produced in rabbit

~1.5 mg/mL

Synonym(s):

Anti-HRCA1, Anti-RCA1, Anti-VHL1, Anti-von Hippel-Lindau tumor suppressor

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41
Product SAB4200285 is not currently sold in your country. Contact Technical Service

biological source

rabbit

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

antigen ~27 kDa

species reactivity

human

concentration

~1.5 mg/mL

technique(s)

immunohistochemistry: 10-20 μg/mL using formalin-fixed paraffin-embedded human kidney.
indirect immunofluorescence: 5-10 μg/mL using HeLa cells.
western blot: 1-2 μg/mL using HEK-293T cells overexpressing human VHL.

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... VHL(7428)

General description

Von Hippel-Lindau (VHL) gene encodes a 213 amino acid polypeptide. VHL interacts stably with proteins like Fibronectin, Elongin B and Elongin C, Cullin-2 and Rbx1. pVHL is shuttled from nucleus to cytoplasm depending on the transcription manner. VHL tumor suppressor gene is located on human chromosome 3p25.3. VHL disease is an autosomal dominant disorder.

Immunogen

synthetic peptide corresponding to a sequence located at the C-terminus of human VHL, conjugated to KLH. The corresponding sequence is identical in human VHL isoform 2.

Biochem/physiol Actions

Von Hippel-Lindau (VHL) is known to regulate cellular metabolism by modulating hypoxia-inducible factor (HIF-1) proteins. VHL protein (pVHL) is used as a marker for proliferative lesions. VHL disease is a hereditary cancer syndrome, which is characterized by vascular tumors. It is caused by the inactivation of VHL protein. Expression of VHL gene is deregulated in papillary thyroid cancers (PTC). Mutations in this gene causes sporadic clear cell renal carcinomas as well as von Hippel-Lindau disease, which is characterised by hemangioblastoma.

Physical form

Solution in 0.01 M phos­phate buffered saline, pH 7.4, containing 15 mM sodium azide.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class

10 - Combustible liquids

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Ubiquitination of hypoxia-inducible factor requires direct binding to the beta-domain of the von Hippel-Lindau protein
Ohh, Michael et al.
Nature Cell Biology, 2(7), 423-423 (2000)
Von Hippel-Lindau disease: a single gene, several hereditary tumors
Crespigio, J et al.
Journal of Endocrinological Investigation, 41(1), 21-31 (2018)
Expression of von Hippel-Lindau tumor suppressor protein (pVHL) characteristic of tongue cancer and proliferative lesions in tongue epithelium
Hasegawa H, et al.
BMC Cancer, 17(1), 381-381 (2017)
Haemangioblastoma of the central nervous system in von Hippel-Lindau disease. French VHL Study Group.
Richard, S et al.
Journal of Internal Medicine, 243(6), 547-553 (1998)
1H NMR metabolomics analysis of renal cell carcinoma cells: Effect of VHL inactivation on metabolism
Cuperlovic-Culf et al.
International Journal of Cancer. Journal International Du Cancer, 138(10), 2439-2449 (2016)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service