Skip to Content
MilliporeSigma
All Photos(3)

Key Documents

SAB4200269

Sigma-Aldrich

Anti-Tensin 2 (N-terminal) antibody produced in rabbit

enhanced validation

~1.5 mg/mL, affinity isolated antibody

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

rabbit

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

antigen ~160 kDa

species reactivity

human

enhanced validation

recombinant expression
Learn more about Antibody Enhanced Validation

concentration

~1.5 mg/mL

technique(s)

indirect immunofluorescence: 1-2 μg/mL using NIH3T3 cells over expressing human tensin2
western blot: 1.5-3.0 μg/mL using lysates of HEK-293T cells over expressing human tensin2

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... TENC1(23371)
mouse ... Tenc1(209039)
rat ... Tenc1(315326)

General description

Tensin 2 belongs to the tensin family. Tensins are multidomain proteins that are characterized with homologous C1, protein phosphatases (PTPases), C2, Src homology 2 (SH2) domains and pTyr-binding (PTB) domains. Tensin 2 mRNA is highly expressed in heart, kidney, skeletal muscle and liver.

Immunogen

synthetic peptide located near the N-terminus human tensin 2, conjugated to KLH. The corresponding sequence is identical in human tensin 2 isoforms 2 and 3, and in rat and mouse tensin 2.

Application

Anti-Tensin 2 (N-terminal) antibody produced in rabbit has been used in various immunochemical techniques including immunoblotting and immunofluorescence.

Biochem/physiol Actions

Tensins are a family of focal adhesion proteins that link between the extracellular matrix (ECM) and the cytoskeleton via integrins, and therefore they might play a role in regulating cell shape and motility. Tensin 2 has been shown to be largely absent in a panel of human cancer cell lines and is down-regulated in human kidney cancer. Tensin 2 variant 3 also contributes to the aggressive tumor behavior in human hepatocellular carcinoma (HCC). The protein through its binding domains interacts with several binding partners and with the tumor suppressor gene deleted in liver cancer 1 (DLC1) to regulate DLC1 tumor suppressor activity. In addition, tensin 2 also stops Akt signaling and cell survival. It reduces intracellular PtdIns-3-4-5-P3 levels at the plasma membrane, indicating that in addition to regulating cytoskeletal dynamics, it influences phosphoinositide-Akt signaling via protein phosphatases (PTPases)domain.

Physical form

solution in 0.01 M phos­phate buffered saline, pH 7.4, containing 15 mM sodium azide.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class

10 - Combustible liquids

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Tensin2 variant 3 is associated with aggressive tumor behavior in human hepatocellular carcinoma
Yam JW, et al.
Hepatology, 44(4), 881-890 (2006)
Tensins: a new switch in cell migration
Mouneimne G and Brugge JS
Developmental Cell, 13(3), 317-319 (2007)
Deleted in liver cancer 1 (DLC1) utilizes a novel binding site for Tensin2 PTB domain interaction and is required for tumor-suppressive function
Chan LK, et al.
PLoS ONE, 4(5), e5572-e5572 (2009)
Tensin2 reduces intracellular phosphatidylinositol 3, 4, 5-trisphosphate levels at the plasma membrane
Hafizi S, et al.
Biochemical and Biophysical Research Communications, 399(3), 396-401 (2010)
Individual domains of Tensin2 exhibit distinct subcellular localizations and migratory effects
Hafizi S, et al.
The International Journal of Biochemistry & Cell Biology, 42(1), 52-61 (2010)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service