Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

SAB4200262

Sigma-Aldrich

Anti-PLEKHA7 (C-terminal) antibody produced in rabbit

~1.0 mg/mL, affinity isolated antibody

Synonym(s):

Anti-Pleckstrin homology domain-containing family A member 7

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

rabbit

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

antigen ~130 kDa

species reactivity

human

concentration

~1.0 mg/mL

technique(s)

immunoprecipitation (IP): 2.5-5.0 μg using lysates of HEK-293T cells overexpressing human PLEKHA7
western blot: 0.5-1.0 μg/mL using lysates of HEK- 293T cells overexpressing human PLEKHA7

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

General description

Pleckstrin homology domain-containing family A, member 7 (PLEKHA7) protein contains WW, pleckstrin homology (PH) and coiled-coil (CC) domains. PLEKHA7 protein is a part of cadherin-based protein complex in mammalian epithelial cells. The PLEKHA7 gene is mapped to human chromosome 11p15.2-p15.1.

Specificity

Anti-PLEKHA7 (C-terminal) recognizes human PLEKHA7.

Immunogen

synthetic peptide corresponding to the C- terminal region of human PLEKHA7 conjugated to KLH. The corresponding sequence differs by a single amino acid in mouse.

Application

Anti-PLEKHA7 (C-terminal) antibody produced in rabbit is suitable for immunoblotting and immunoprecipitation.

Biochem/physiol Actions

Pleckstrin homology domain-containing family A, member 7 (PLEKHA7) protein via the cadherin-based protein complex is involved in anchoring the microtubule minus ends to the zonula adherens (ZA). This is essential for the biogenesis of specialized junction. PLEKHA7 binds directly to E-cadherin and cadherin associated protein p120-catenin complexes and appears to be necessary for recruiting NEZHA to the ZA.

Physical form

Solution in 0.01 M phos­phate buffered saline, pH 7.4, containing 15 mM sodium azide.

Storage and Stability

For continuous use, store at 2-8 °C for up to one month. For extended storage, freeze in working aliquots. Repeated freezing and thawing, or storage in “frost-free” freezers,is not recommended. If slight turbidity occurs upon prolonged storage, clarify the solution by centrifugation before use. Working dilutions shouldbe discarded if not used within 12 hours.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class

10 - Combustible liquids

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Catalog of 5?fusion partners in ALK+ NSCLC circa 2020
Ou SHI, et al.
Journal of Chemical and Pharmaceutical Sciences , 100015 null
Wenxiang Meng et al.
Cell, 135(5), 948-959 (2008-12-02)
Epithelial cells contain noncentrosomal microtubules (MTs), whose minus ends are oriented apically. In contrast with the well-known interactions of the minus ends with the centrosome, little is known about the termination site of the noncentrosomal minus ends. Here we show
Anna Akhmanova et al.
Cell, 135(5), 791-793 (2008-12-02)
The zonula adherens (ZA) is a specialized cadherin-based structure found at the contacts between epithelial cells. Meng et al. (2008) now identify a protein complex containing the microtubule minus-end-binding protein Nezha, which provides a critical link between microtubules and cadherins

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service