Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

SAB4200260

Sigma-Aldrich

Anti-FIP1L1 (C-terminal) antibody produced in rabbit

~1.0 mg/mL, affinity isolated antibody

Synonym(s):

Anti-FIP1, Anti-RHE, Anti-hFip1, Anti-pre-mRNA 3’end processing factor FIP1

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

rabbit

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

antigen ~70 kDa

species reactivity

human

concentration

~1.0 mg/mL

technique(s)

indirect immunofluorescence: 2.5-5 μg/mL using paraformaldehyde fixed NIH-3T3 cells
western blot: 2-4 μg/mL using lysates of NIH-3T3 cells

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

General description

The factor interacting with poly(a) polymerase α (PAPOLA) and cleavage and polyadenylation specificity factor 1 (CPSF1) (FIP1L1) is a subunit of CPSF complex. FIP1L1 contains an acidic N-terminal region, a proline-rich domain and a C-terminal region of alternating arginines and aspartates (RD domain). The FIP1L1 gene is mapped to human chromosome 4q12.

Specificity

Anti-FIP1L1 (C-terminal) recognizes human FIP1L1.

Immunogen

synthetic peptide corresponding to a sequence at the C- terminal region of human FIP1L1 conjugated to KLH. The corresponding sequence is identical in mouse and rat.

Application

Anti-FIP1L1 (C-terminal) antibody produced in rabbit is suitable for immunoblotting and immunofluorescence.

Biochem/physiol Actions

The factor interacting with poly(a) polymerase α (PAPOLA) and cleavage and polyadenylation specificity factor 1 (CPSF1) (FIP1L1) is known to promote the U-rich sequence element mediated poly(A) polymerase (PAP) action. Genetic rearrangements that result in a FIP1L1- platelet-derived growth factor receptor A (PDGFRA) fusion gene have been identified in patients with hypereosinophilic syndrome/chronic eosinophilic leukemia (CEL) and in systemic mast cell disease.

Physical form

Solution in 0.01 M phos­phate buffered saline, pH 7.4, containing 15 mM sodium azide.

Storage and Stability

For continuous use, store at 2-8 °C for up toone month. For extended storage, freeze in working aliquots. Repeated freezing and thawing, or storage in “frost-free” freezers, is not recommended. If slight turbidity occurs upon prolonged storage, clarify the solution by centrifugation before use. Working dilutions should be discarded if not used within 12 hours.

Disclaimer

Unless otherwise stated in our catalog, our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class

10 - Combustible liquids

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Yoshiyuki Yamada et al.
International archives of allergy and immunology, 152 Suppl 1, 101-105 (2010-06-11)
Since the identification of the FIP1L1/PDGFRA fusion gene as a pathogenic cause of the hypereosinophilic syndrome (HES), the importance of the molecular classification of HES leading to the diagnosis of chronic eosinophilic leukemia (CEL) has been recognized. As a result
Isabelle Kaufmann et al.
The EMBO journal, 23(3), 616-626 (2004-01-30)
In mammals, polyadenylation of mRNA precursors (pre-mRNAs) by poly(A) polymerase (PAP) depends on cleavage and polyadenylation specificity factor (CPSF). CPSF is a multisubunit complex that binds to the canonical AAUAAA hexamer and to U-rich upstream sequence elements on the pre-mRNA

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service