Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

SAB4200254

Sigma-Aldrich

Anti-SIRT6 antibody, Mouse monoclonal

clone SIRT6-4, purified from hybridoma cell culture

Synonym(s):

Monoclonal Anti-NAD-dependent deacetylase sirtuin-6, Monoclonal Anti-SIR2-like protein 6, Monoclonal Anti-SIR2L6, Monoclonal Anti-sirtuin 6

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

mouse

conjugate

unconjugated

antibody form

purified from hybridoma cell culture

antibody product type

primary antibodies

clone

SIRT6-4, monoclonal

form

buffered aqueous solution

mol wt

antigen ~40 kDa

species reactivity

human

concentration

~1.0 mg/mL

technique(s)

indirect immunofluorescence: suitable
western blot: 1-2 μg/mL using whole extracts of HEK-293T cells overexpressing human SIRT6.

isotype

IgG2b

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... SIRT6(51548)
mouse ... Sirt6(50721)

General description

Monoclonal Anti-SIRT6 (mouse IgG2b isotype) is derived from the hybridoma SIRT6-4 produced by the fusion of mouse myeloma cells and splenocytes from BALB/c mice immunized with a synthetic peptide corresponding to the N-terminal region of human Sirt6 conjugated to KLH. Sirtuin 6 (Sirt6) belongs to the mammalian Sir2 gene family. It is a nuclear, chromatin-associated protein. Sirt6 is broadly expressed with the highest levels in muscle, brain and heart.

Immunogen

synthetic peptide corresponding to the N-terminal region of human Sirt6 (GeneID: 51548), conjugated to KLH.

Application

Monoclonal Anti-SIRT6 antibody produced in mouse has been used in immunoblotting.

Biochem/physiol Actions

Sirtuin 6 (Sirt6) plays a key role in DNA repair and maintenance of genomic stability in cells. Sirt6 is enzymatically inactive as a Nicotinamide adenine dinucleotide (NAD+) -dependent protein deacetylase but displays auto-adenosine triphosphate (ADP)-ribosyl transferase activity. Loss of Sirt6 leads to the development of an acute degenerative aging-like phenotype.

Physical form

Solution in 0.01 M phosphate buffered saline, pH 7.4, containing 15 mM sodium azide.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class

12 - Non Combustible Liquids

wgk_germany

WGK 1

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Ahmet Can Timucin et al.
PloS one, 11(8), e0161494-e0161494 (2016-08-19)
SIRT6 is a protein deacetylase, involved in various intracellular processes including suppression of glycolysis and DNA repair. Aldose Reductase (AR), first enzyme of polyol pathway, was proposed to be indirectly associated to these SIRT6 linked processes. Despite these associations, presence
SIRT6 is a positive regulator of aldose reductase expression in U937 and HeLa cells under osmotic stress: in vitro and in silico insights
Timucin AC and Basaga H
PLoS ONE, 11(8), e0161494-e0161494 (2016)
Genomic instability and aging-like phenotype in the absence of mammalian SIRT6
Mostoslavsky R, et al.
Cell, 124(2), 315-329 (2006)
Mouse Sir2 homolog SIRT6 is a nuclear ADP-ribosyltransferase
Liszt G, et al.
The Journal of biological chemistry, 280(22), 21313-21320 (2005)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service