Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

SAB4200246

Sigma-Aldrich

Anti-Ambra1 antibody produced in rabbit

~1.0 mg/mL, affinity isolated antibody

Synonym(s):

Anti-activating molecule in BECN1-regulated autophagy protein 1, Anti-autophagy/beclin 1 regulator 1

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203

biological source

rabbit

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

~130 kDa

species reactivity

human

concentration

~1.0 mg/mL

technique(s)

western blot: 2-4 μg/mL using HEK-293T cells overexpressing human Ambra1.

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

Gene Information

General description

Activating molecule in Beclin 1-regulated autophagy (Ambra1) is a WD40-containing protein.[1] It is mainly expressed in the neuroepithelium during early neurogenesis. Ambra1 is highly conserved in vertebrates.[1] The gene is located on human chromosome 11p11.2.[2]

Immunogen

synthetic peptide corresponding to the N-terminal region of human Ambra1, conjugated to KLH. The corresponding sequence is identical in mouse and rat Ambra1.

Application

Anti-Ambra1 antibody produced in rabbit has been used in immunoblotting.

Biochem/physiol Actions

Activating molecule in Beclin 1-regulated autophagy (Ambra1) is a positive regulator of Beclin1-regulated autophagy. It is involved in the development of the nervous system. Upon autophagic stimuli, Ambra1 binds to Beclin1 and promotes the interaction of Beclin1 with its target phosphatidylinositol 3-kinase VPS34, thereby inducing autophagy. Functional deficiency of Ambra1 in mouse embryos is associated with severe neural tube defects such as autophagy impairment, unbalanced cell proliferation, accumulation of ubiquitinated proteins and excessive apoptosis.[1]

Physical form

Solution in 0.01 M phos­phate buffered saline, pH 7.4, containing 15 mM sodium azide.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class

10 - Combustible liquids

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Ambra1 regulates autophagy and development of the nervous system
Fimia GM, et al.
Nature, 447(7148), 1121-1121 (2007)
Heterozygous ambra1 deficiency in mice: a genetic trait with autism-like behavior restricted to the female gender
Dere E, et al.
Frontiers in Behavioral Neuroscience, 8, 181-181 (2014)
Takeo Nitta et al.
International journal of clinical and experimental pathology, 7(8), 4913-4921 (2014-09-10)
The role of autophagy in cholangiocarcinoma is poorly understood. This study investigated its involvement in cholangiocarcinoma, focusing on carcinoma cell invasion and prognostic significance using cholangiocarcinoma cell lines, CCKS1 and HuCCT1, and human tissues of hilar and extrahepatic cholangiocarcinoma. Nutrient

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service