Monoclonal Anti-Leucine carboxyl methyltransferase 1 (LCMT1) (mouse IgG1 isotype) is derived from the hybridoma 4A4 produced by the fusion of mouse myeloma cells and splenocytes from the immunized BALB/c mice.
Specificity
Monoclonal Anti-Leucine carboxyl methyltransferase 1 (LCMT1) recognizes mouse and rat LCMT1.
Immunogen
mouse LCMT1 fusion protein.
Application
Anti-Leucine carboxyl methyltransferase 1 antibody, Mouse monoclonal is suitable for immunoblotting.
Biochem/physiol Actions
In mammalian cells, leucine carboxyl methyltransferase 1 (LCMT1), also known as protein phosphatase 2A (PP2A) methyltransferase 1 (PPMT1) is responsible for the methylation of the carboxyl group of the C-terminal leucine residue (Leu309) of the catalytic subunit of PP2A. Methylation regulates PP2A by modulating the incorporation of different B-type subunits into PP2A holoenzymes, thereby affecting subcellular targeting and substrate specificity. LCMT1 is crucial for normal mitosis and cell survival, and for embryonic development in mice. Downregulation of LCMT1 and PP2A methylation are observed in Alzheimer′s disease patients and hyperhomocysteinemic mice.
Physical form
Solution in 0.01 M phosphate buffered saline, pH 7.4, containing 15 mM sodium azide.
Storage and Stability
For continuous use, store at 2-8°C for up to one month. For extended storage, freeze at -20°C in working aliquots. Repeated freezing and thawing,or storage in “frost-free” freezers,is not recommended. If slight turbidity occurs upon prolonged storage, clarify the solution by centrifugation before use. Working dilution samples should be discardedif not used within 12 hours.
Disclaimer
Unless otherwise stated in our catalog, our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.
The Journal of biological chemistry, 282(42), 30974-30984 (2007-08-29)
Protein phosphatase 2A (PP2A) is a multifunctional phosphatase that plays important roles in many cellular processes including regulation of cell cycle and apoptosis. Because PP2A is involved in so many diverse processes, it is highly regulated by both non-covalent and
The Journal of neuroscience : the official journal of the Society for Neuroscience, 28(45), 11477-11487 (2008-11-07)
Altered folate homeostasis is associated with many clinical and pathological manifestations in the CNS. Notably, folate-mediated one-carbon metabolism is essential for methyltransferase-dependent cellular methylation reactions. Biogenesis of protein phosphatase 2A (PP2A) holoenzyme containing the regulatory B(alpha) subunit, a major brain
Questions
Reviews
★★★★★ No rating value
Active Filters
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.