Skip to Content
MilliporeSigma
All Photos(3)

Key Documents

SAB4200238

Sigma-Aldrich

Anti-LAP2α antibody, Mouse monoclonal

clone 3A3, purified from hybridoma cell culture

Synonym(s):

Anti-LEM domain containing 4, Anti-LEMD4, Anti-Lamina-associated polypeptide 2, Anti-TMPO, Anti-TP α, Anti-Thymopoietin isoform α, Anti-Thymopoietin-related peptide isoform α

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352203
NACRES:
NA.41

biological source

mouse

conjugate

unconjugated

antibody form

purified from hybridoma cell culture

antibody product type

primary antibodies

clone

3A3, monoclonal

form

buffered aqueous solution

mol wt

antigen ~80 kDa

species reactivity

human, bovine

concentration

~1.0 mg/mL

technique(s)

immunoprecipitation (IP): suitable
western blot: 1-2 μg/mL using whole extracts of human HeLa cells.

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... TMPO(7112)

General description

Monoclonal Anti-LAP2α (mouse IgG1 isotype) is derived from the hybridoma 3A3 produced by the fusion of mouse myeloma cells and splenocytes from BALB/c mice immunized with a peptide. Lamina-associated polypeptide 2, isoform α (LAP2α), also called thymopoietin isoform α, is encoded by the gene mapped to human chromosome 12q23.1. LAP2α is a LEM (LAP2-Emerin-MAN1) domain protein. that resides in the nucleus.

Specificity

Monoclonal Anti-LAP2α recognizes human and bovine LAP2α.

Immunogen

peptide corresponding to the C-terminus of human LAP2α, conjugated to KLH.

Application

Monoclonal Anti-LAP2α antibody produced in mouse may be used in immunoblotting and immunoprecipitation.

Biochem/physiol Actions

Lamina-associated polypeptide 2 α (LAP2 α) may play a role in the assembly of the nuclear lamina, and thus helps in maintaining the structural organization of the nuclear envelope. It may also facilitate the binding of lamin A to the inner nuclear membrane. Mutation in this gene leads to the development of dilated cardiomyopathy.

Physical form

Solution in 0.01 M phosphate buffered saline, pH 7.4, containing 15 mM sodium azide.

Storage and Stability

For continuous use, store at 2-8°C for up to one month. For extended storage, freeze at -20oC in working aliquots. Repeated freezing and thawing,or storage in “frost-free” freezers,is not recommended. If slight turbidity occurs upon prolonged storage, clarify the solution by centrifugation before use. Working dilution samples should be discardedif not used within 12 hours.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class

12 - Non Combustible Liquids

wgk_germany

WGK 1

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Nuclear envelopathies: a complex LINC between nuclear envelope and pathology
Janin A, et al.
Orphanet Journal of Rare Diseases, 12(1), 147-147 (2017)
Sudden cardiac death in young athletes: Literature review of molecular basis
Mazzaccara C,et al.
Carcinogenesis, 10(1) (2020)
Proteins that bind A-type lamins: integrating isolated clues
Zastrow MS, et al.
Journal of Cell Science, 117(7), 979-987 (2004)
Lamins in the nuclear interior- life outside the lamina
Naetar N, et al.
Journal of Cell Science, 130(13), 2087-2096 (2017)
Anna Kychygina et al.
Scientific reports, 11(1), 13195-13195 (2021-06-25)
Chromatin organization within the nuclear volume is essential to regulate many aspects of its function and to safeguard its integrity. A key player in this spatial scattering of chromosomes is the nuclear envelope (NE). The NE tethers large chromatin domains

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service