Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

SAB4200229

Sigma-Aldrich

Anti-CD3ζ chain antibody, Mouse monoclonal

clone TCR48, purified from hybridoma cell culture

Synonym(s):

Anti-CD247 antigen, Anti-CD3-eta, Anti-Cd3, Anti-Cd3h, Anti-T3z, Anti-TCRZ

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352203
NACRES:
NA.41

biological source

mouse

conjugate

unconjugated

antibody form

purified from hybridoma cell culture

antibody product type

primary antibodies

clone

TCR48, monoclonal

form

buffered aqueous solution

mol wt

antigen ~16 kDa

species reactivity

human, mouse

concentration

~1.0 mg/mL

technique(s)

immunoprecipitation (IP): suitable
western blot: 2.0-4.0 μg/mL using Jurkat or EL4 cell extracts

isotype

IgG1

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

mouse ... Cd247(12503)

General description

Monoclonal Anti- CD3ζ chain (mouse IgG1 isotype) is derived from the hybridoma TCR-48 produced by the fusion of mouse myeloma cells and splenocytes from BALB/c mice immunized with a synthetic peptide corresponding to a fragment of mouse CD3ζ chain conjugated to KLH. CD3ζ is a 16-kDa transmembrane protein, which consists of a short extracellular domain and a long intracellular domain including immunoreceptor tyrosine-based activation motif (ITAM). It is a subunit of T cell receptor chains (TCR), which is expressed by T cells and natural killer (NK) cells.

Application

Monoclonal Anti-CD3ζ chain antibody produced in mouse has been used in immunoblotting and immunoprecipitation.

Biochem/physiol Actions

CD3ζ is required for T cell receptor chains (TCR) complex assembly and expression. It is involved in immunity, neuronal development and synaptic plasticity. Downregulation of TCR ζ-chain expression and impairment of T-cell function is associated with systemic lupus erythematosus (SLE), rheumatoid arthritis, cancer and infectious diseases. Altered expression and function of CD3ζ in T cells is linked to hematological diseases.

Physical form

Solution in 0.01M phosphate buffered saline pH 7.4, containing 15 mM sodium azide.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class

12 - Non Combustible Liquids

wgk_germany

WGK 1

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

T cell CD3zeta deficiency enables multiorgan tissue inflammation
Deng GM, et al.
Journal of immunology (Baltimore, Md. : 1950), 191(7), 3563-3567 (2013)
Cutting edge: CD3 ITAM diversity is required for optimal TCR signaling and thymocyte development
Bettini ML, et al.
Journal of Immunology, 199(5), 1555-1560 (2017)
Alterations in the expression pattern of TCR zeta chain in T cells from patients with hematological diseases
Li Y
Hematology, 13(5), 267-275 (2008)
Antoine Louveau et al.
The Journal of neuroscience : the official journal of the Society for Neuroscience, 33(47), 18672-18685 (2013-11-22)
The immunoreceptor-associated protein CD3ζ is known for its role in immunity and has also been implicated in neuronal development and synaptic plasticity. However, the mechanism by which CD3ζ regulates synaptic transmission remains unclear. In this study, we showed that mice

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service