Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

SAB4200220

Sigma-Aldrich

Monoclonal Anti-Rab25 antibody produced in mouse

~1.0 mg/mL, clone Rab25-5, purified immunoglobulin

Synonym(s):

Anti-CATX-8, Anti-RAB11C, Anti-Ras-related protein Rab-25

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

mouse

conjugate

unconjugated

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

Rab25-5, monoclonal

form

buffered aqueous solution

mol wt

antigen ~25 kDa

species reactivity

human

concentration

~1.0 mg/mL

technique(s)

western blot: 2-4 μg/mL using whole extracts of HEK-293T cells over-expressing human Rab25

isotype

IgG1

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... RAB25(57111)

Looking for similar products? Visit Product Comparison Guide

General description

Monoclonal Anti-Rab25 (mouse IgG1 isotype) is derived from the hybridoma Rab25-5 produced by the fusion of mouse myeloma cells and splenocytes from the immunized BALB/c mice. Ras-related protein Rab 25 (Rab25) is a member of the Rab family of small guanosine triphosphatases (GTPases) restricted to an epithelial distribution. The Rab25 belongs to the Rab11 subfamily. The RAB25 gene is mapped to human chromosome 1q22.

Specificity

Monoclonal Anti-Rab25 recognizes human Rab25.

Application

Monoclonal Anti-Rab25 antibody produced in mouse is suitable for immunoblotting.

Biochem/physiol Actions

Ras-related protein Rab (Rab) guanosine triphosphatase (GTPases) are central regulators of membrane trafficking between the different subcellular compartments of the eukaryotic cell. Their regulatory capacity is determined by their ability to cycle between the guanosine diphosphate (GDP)-bound inactive and guanosine triphosphate (GTP)-bound active states. Conversion from one state to the other is regulated by GDP/GTP exchange factors (GEFs), GDP dissociation inhibitors (GDIs), and GTPase-activating proteins (GAPs). Rab protein gets activated by associating with the intracellular membranes and also recruits the downstream effector proteins to the cytoplasmic surface of a subcellular compartment. Rab GTPases regulate the effector proteins mediated vesicle formation, membrane fusion, and actin- and tubulin-dependent vesicle movement. The RAB25 gene expression is associated with the progression of breast, ovarian and prostate cancer cells and also mammary cell lines.

Physical form

Solution in 0.01 M phosphate buffered saline, pH 7.4, containing 15 mM sodium azide.

Storage and Stability

For continuous use, store at 2–8 °C for up to one month. For extended storage, freeze at –20 °C in working aliquots. Repeated freezing and thawing, or storage in “frost-free” freezers, is not recommended. If slight turbidity occurs upon prolonged storage, clarify the solution by centrifugation before use. Working dilution samples should be discarded if not used within 12 hours.

Disclaimer

Unless otherwise stated in our catalog, our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class

10 - Combustible liquids

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Regulators and Effectors of Small GTPases
Methods in Enzymology, 3-553 null
Oligomerization of rab/effector complexes in the regulation of vesicle trafficking
Progress in Molecular Biology and Translational Science, 579-614 null
Jae Hong No et al.
Anticancer research, 32(8), 3467-3471 (2012-07-31)
The use of cell-free DNA (cfDNA) as a non-invasive biomarker has been evaluated in many types of cancer. This study investigated the prognostic significance of cfDNA level for ovarian cancer. Preoperative sera of 36 patients with ovarian cancer and of
Bassam R Ali et al.
Journal of cell science, 117(Pt 26), 6401-6412 (2004-11-25)
Small GTPases of the Rab family are key regulators of membrane trafficking. Each Rab shows a characteristic subcellular distribution, and may serve as an important determinant of organelle identity. The molecular mechanisms responsible for targeting Rabs to specific intracellular compartments
Y Takai et al.
Physiological reviews, 81(1), 153-208 (2001-01-12)
Small GTP-binding proteins (G proteins) exist in eukaryotes from yeast to human and constitute a superfamily consisting of more than 100 members. This superfamily is structurally classified into at least five families: the Ras, Rho, Rab, Sar1/Arf, and Ran families.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service