Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

SAB4200213

Sigma-Aldrich

Monoclonal Anti-Syntrophin antibody produced in mouse

~1.0 mg/mL, clone 1351, purified immunoglobulin

Synonym(s):

Anti-Pro-TGF-α cytoplasmic domain-interacting protein 1, Anti-SNTA1, Anti-Syntrophin 1, SNT159 kDa dystrophin-associated protein A1 acidic component 1, Anti-Syntrophin, α1

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203

biological source

mouse

conjugate

unconjugated

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

1351, monoclonal

form

buffered aqueous solution

mol wt

antigen 58 kDa

species reactivity

rat, chicken, human, Xenopus, frog, mouse

concentration

~1.0 mg/mL

technique(s)

electron microscopy: suitable
immunoprecipitation (IP): suitable
indirect immunofluorescence: suitable
western blot: 0.5-1.0 μg/mL using whole extracts of rat heart or L8 cells.

isotype

IgG1

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

Gene Information

human ... SNTA1(6640)
mouse ... Snta1(20648)
rat ... Snta1(362242)

Immunogen

Torpedo electric organ membranes.

Physical form

Solution in 0.01 M phosphate buffered saline, pH 7.4, containing 15 mM sodium azide.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class

12 - Non Combustible Liquids

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

S C Froehner et al.
The Journal of cell biology, 104(6), 1633-1646 (1987-06-01)
In the study of proteins that may participate in the events responsible for organization of macromolecules in the postsynaptic membrane, we have used a mAb to an Mr 58,000 protein (58K protein) found in purified acetylcholine receptor (AChR)-enriched membranes from
Kristina Eisinger et al.
Analytical biochemistry, 484, 99-101 (2015-06-17)
Alpha-syntrophin (SNTA) is an adaptor protein that regulates several signaling pathways. To analyze expression of SNTA immunoblot assays must be performed. Here, the specificity of four commercially available SNTA antibodies has been evaluated in immunoblot experiments using liver tissues of
Tugba Can et al.
Proteome science, 12, 25-25 (2014-07-30)
The myotendinous junction is a specialized structure of the muscle fibre enriched in mechanosensing complexes, including costameric proteins and core elements of the z-disc. Here, laser capture microdissection was applied to purify membrane regions from the myotendinous junctions of mouse
Kristy Swiderski et al.
Disease models & mechanisms, 17(4) (2024-04-11)
Duchenne muscular dystrophy (DMD) is a devastating monogenic skeletal muscle-wasting disorder. Although many pharmacological and genetic interventions have been reported in preclinical studies, few have progressed to clinical trials with meaningful benefit. Identifying therapeutic potential can be limited by availability

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service