Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

SAB4200209

Sigma-Aldrich

Anti-PGC-1α antibody produced in rabbit

~1.5 mg/mL, affinity isolated antibody

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352203

biological source

rabbit

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

antigen ~120 kDa

species reactivity

human

concentration

~1.5 mg/mL

technique(s)

western blot: 1-2 μg/mL using HEK-293T cells overexpressing human PGC-1α

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

Gene Information

Application

Applications in which this antibody has been used successfully, and the associated peer-reviewed papers, are given below.
Western Blotting (1 paper)

Physical form

Solution in 0.01 M phosphate buffered saline, pH 7.4, containing 15 mM sodium azide.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class

12 - Non Combustible Liquids

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Erin J Stephenson et al.
American journal of physiology. Endocrinology and metabolism, 310(11), E1003-E1015 (2016-04-28)
We have investigated the effects of in utero exposure to environmentally persistent free radicals (EPFRs) on growth, metabolism, energy utilization, and skeletal muscle mitochondria in a mouse model of diet-induced obesity. Pregnant mice were treated with laboratory-generated, combustion-derived particular matter
Yiping Zhang et al.
Evidence-based complementary and alternative medicine : eCAM, 2015, 649568-649568 (2015-07-23)
The lack of effective treatment for liver cirrhosis and hepatocellular carcinomas imposes serious challenges to the healthcare system. Here, we investigated the efficacy and mechanism of liquiritigenin involved in preventing or retarding the progression of liver diseases in a rat
Mithra Sudha Mohan et al.
Physiology international, 110(1), 1-18 (2023-02-28)
Hypothermic conditions enhance the incidence of cardiovascular diseases due to increased blood pressure. Cold-induced adaptive thermogenesis increased mitochondrial biogenesis and function in skeletal muscles and adipocytes. Here, we studied the effect of intermittent cold exposure on the regulators of cardiac

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service