Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

SAB4200204

Sigma-Aldrich

Anti-Tensin 3 antibody produced in rabbit

~1.5 mg/mL, affinity isolated antibody

Synonym(s):

Anti-TEM6, Anti-TNS3, Anti-Tensin-like SH2 domain-containing protein 1, Anti-Tumor endothelial marker 6

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

rabbit

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

antigen ~180 kDa

species reactivity

human

concentration

~1.5 mg/mL

technique(s)

western blot: 1.5-3.0 μg/mL using HEK-293T cell lysates over expressing human tensin 3.

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

Gene Information

human ... TNS3(64759)

Physical form

Solution in 0.01 M phos­phate buffered saline, pH 7.4, containing 15 mM sodium azide.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class

10 - Combustible liquids

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Brajendra K Tripathi et al.
The Journal of cell biology, 207(5), 627-642 (2014-12-03)
DLC1 is a tumor suppressor protein whose full activity depends on its presence at focal adhesions, its Rho-GTPase activating protein (Rho-GAP) function, and its ability to bind several ligands, including tensin and talin. However, the mechanisms that regulate and coordinate

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service