Skip to Content
MilliporeSigma
All Photos(3)

Key Documents

SAB4200200

Sigma-Aldrich

Anti-LMOD1 antibody produced in rabbit

~1.0 mg/mL, affinity isolated antibody

Synonym(s):

Anti-64 kDa autoantigen 1D3, Anti-64 kDa autoantigen D1, Anti-Hashimoto thyroiditis autoantigen, 64-KD, Anti-Leiomodin 1 (smooth muscle), Anti-SM-LMOD, 64 kDa autoantigen 1D, Anti-Thyroid and eye muscle autoantigen D1 (64kD), Anti-Thyroid-associated ophthalmopathy autoantigen

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

rabbit

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

antigen ~72 kDa

species reactivity

human, mouse, rat

packaging

antibody small pack of 25 μL

concentration

~1.0 mg/mL

technique(s)

immunohistochemistry: 10-20 μg/mL using heat-retrieved formalin-fixed, paraffin-embedded human and mouse intestine sections and biotin / ExtrAvidin® Peroxidase staining system
western blot: 2.5-5.0 μg/mL using whole extracts of rat H9C2 cells

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... LMOD1(25802)
mouse ... Lmod1(93689)
rat ... Lmod1(304816)

Physical form

Solution in 0.01 M phos­phate buffered saline, pH 7.4, containing 15 mM sodium azide.

Legal Information

ExtrAvidin is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class

10 - Combustible liquids

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service