Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

SAB4200196

Sigma-Aldrich

Anti-C/EBPβ antibody, Rat monoclonal

clone 7H5, purified from hybridoma cell culture

Synonym(s):

Anti-CCAAT/enhancer binding protein (C/EBP), beta, Anti-CEBPB, Anti-CRP2, Anti-IL6DBP, Anti-LAP, Anti-NF-IL6, Anti-TCF5, Monoclonal Anti-C/EBPβ antibody produced in rat

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352203
NACRES:
NA.41

biological source

rat

conjugate

unconjugated

antibody form

purified from hybridoma cell culture

antibody product type

primary antibodies

clone

7H5, monoclonal

form

buffered aqueous solution

mol wt

~37 kDa

species reactivity

mouse

packaging

antibody small pack of 25 μL

concentration

~1.0 mg/mL

technique(s)

immunocytochemistry: suitable
western blot: 2.0-4.0 μg/mL using L929 or 3T3 cell extracts

isotype

IgG2a

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

mouse ... CEBPB(12608)

General description

Monoclonal Anti-C/EBPβ (rat IgG2a isotype) is derived from the hybridoma 7H5 produced by the fusion of mouse myeloma cells (SP2) and splenocytes from rat immunized with a mouse C/EBPβ fusion protein. CCAAT/enhancer binding proteins β (C/EBPβ) belongs to the highly conserved family of C/EBP transcription factors. It possesses a C-terminal basic region-leucine zipper (bZIP) domain, which is essential for DNA binding. C/EBPβ has two central regulatory domains (RDs), RD1 and RD2.

Immunogen

mouse C/EBPβ fusion protein

Application

Monoclonal Anti-C/EBPβ antibody produced in rat has been used in immunoblotting and immunocytochemistry.

Biochem/physiol Actions

CCAAT/enhancer binding proteins β (C/EBPβ) exhibits plasticity with respect to various transcriptional activities it takes part in. It regulates IL-6 (interleukin-6) and IL-6-induced expression of the cytokines IL-1, IL-8, tumor necrosis factor-α (TNFα) and granulocyte colony-stimulating factor (G-CSF). C/EBPβ modulates acute phase response proteins such as α1-acid glycoprotein, α2-microglobulin and C-reactive protein. It is implicated in the development of the mammary gland and , in cell proliferation. C/EBPβ regulates survival in liver and in response to injuries and hormones in lungs. It plays a role in human breast tumorigenesis.

Physical form

Solution in 0.01M phosphate buffered saline, pH 7.4, containing 15 mM sodium azide

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class

10 - Combustible liquids

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

The interferon signaling network and transcription factor C/EBP-beta
Li H, et al.
Cellular & Molecular Immunology, 4(6), 407-407 (2007)
C/EBP transcription factors in the lung epithelium
Cassel TN and Nord M
American Journal of Physiology. Lung Cellular and Molecular Physiology, 285(4), L773-L781 (2003)
CCAAT/enhancer binding proteins in normal mammary development and breast cancer
Zahnow CA
Breast Cancer Research, 4(3), 113-113 (2002)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service