Skip to Content
MilliporeSigma
All Photos(3)

Key Documents

SAB4200168

Sigma-Aldrich

Anti-Importin 4 (N-terminal) antibody produced in rabbit

~1.0 mg/mL, affinity isolated antibody

Synonym(s):

Anti-IPO4, Anti-Ran-binding protein 4, Anti-imp4b, Anti-ranBP4

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

rabbit

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

antigen ~120 kDa

species reactivity

human

concentration

~1.0 mg/mL

technique(s)

immunoprecipitation (IP): 2.5-5.0 μg using HepG2 cell lysates
indirect immunofluorescence: 1.25-2.5 μg/mL using HEK-293T cells overexpressing human Importin 4
western blot: 1-2 μg/mL using HepG2 cell lysates

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... IPO4(79711)

General description

Importin 4 is a member of importin protein family. It is a 119 kDa protein.

Immunogen

synthetic peptide corresponding to a sequence at the N-terminal of human Importin 4

Application

Anti-Importin 4 (N-terminal) antibody produced in rabbit has been used in several immunochemical techniques including immunoblotting, immunoprecipitation and immunofluorescence.

Biochem/physiol Actions

Importins can mediate the nuclear import of a diverse set of protein or RNA cargoes. Importins were implicated in anti-aggregation activity of highly basic cargo like histones and ribosomal proteins and in preventing their undesired interactions with other cellular components. It has been identified as an import factor for ribosomal protein rpS3a, the vitamin D receptor (VDR) and the sperm transition protein transition protein 2 (TP2).

Physical form

Solution in 0.01 M phosphate buffered saline, pH 7.4, containing 15 mM sodium azide

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class

10 - Combustible liquids

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Involvement of importin-4 in the transport of transition protein 2 into the spermatid nucleus
Pradeepa MM, et al.
Molecular and Cellular Biology, 28(13), 4331-4341 (2008)
Importins fulfil a dual function as nuclear import receptors and cytoplasmic chaperones for exposed basic domains
Jakel S, et al.
The Embo Journal, 21(3), 377-386 (2002)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service