Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

SAB4200161

Sigma-Aldrich

Anti-Tensin 2 antibody produced in rabbit

enhanced validation

~0.5 mg/mL, affinity isolated antibody

Synonym(s):

Anti-C1TEN, Anti-TENC1, Anti-TNS2

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

rabbit

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

antigen ~160 kDa

species reactivity

mouse, human, rat

enhanced validation

recombinant expression
Learn more about Antibody Enhanced Validation

concentration

~0.5 mg/mL

technique(s)

western blot: 0.5-1.0 μg/mL using HEK-293T cell lysates overexpressing human tensin 2

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... TENC1(23371)
mouse ... Tenc1(209039)
rat ... Tenc1(315326)

General description

Tensins are a family of focal adhesion proteins that link the extracellular matrix (ECM) and the cytoskeleton via integrins. The tensin family includes four members: tensin 1-4, encoded by different genes. Tensin proteins consists of homologous C1, protein tyrosine phosphatase (PTPase), C2, SH2 and phosphotyrosine-binding domain (PTB) domains. Tensin 2 is very similar to tensin1 at its N- and C-terminus. They have similar actin binding domain, SH2 and PTB domains. Tensin 2 is a binding partner for Axl receptor tyrosine kinase. Tensin 2 mRNA is highly expressed in heart, liver, kidney and skeletal muscle.

Application

Anti-Tensin 2 antibody produced in rabbit has been used in immunoblotting.

Biochem/physiol Actions

Tensins play an important role in regulating cell shape and motility. Abnormal expression of tensins are implicated in various forms of human cancers. Expression of tensin 2 and tensin 3 are markedly reduced in a panel of diverse human cancer cell lines and all four genes are downregulated in human kidney cancer. Tensin 2 induces growth inhibition and apoptosis in hepatocellular carcinoma (HCC) cell lines. Tensin 2 interacts directly with deleted in liver cancer 1 (DLC1), a tumor suppressor protein often deleted and downregulated in HCC cell lines and other types of cancers. Tensin 2 may be important for normal kidney function in vivo.

Physical form

Solution in 0.01 M phos­phate buffered saline, pH 7.4, containing 15 mM sodium azide.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class

10 - Combustible liquids

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

AMPK negatively regulates tensin-dependent integrin activity
Georgiadou M, et al.
The Journal of Cell Biology, 216(4), 1107-1121 (2017)
Interaction of deleted in liver cancer 1 with tensin2 in caveolae and implications in tumor suppression
Yam JWP, et al.
Cancer Research, 66(17), 8367-8372 (2006)
The possible involvement of tensin2 in the expression and extension of nephrin by glomerular podocytes in mice
Kato T, et al.
Biomedical Research, 29(6), 279-287 (2008)
Interaction of Axl receptor tyrosine kinase with C1-TEN, a novel C1 domain-containing protein with homology to tensin
Hafizi S, et al.
Biochemical and biophysical research communications, 299(5), 793-800 (2002)
Tensin1 and a previously undocumented family member, tensin2, positively regulate cell migration
Chen H, et al.
Proceedings of the National Academy of Sciences of the USA, 99(2), 733-738 (2002)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service