Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

SAB4200158

Sigma-Aldrich

Monoclonal Anti- MAGE-11

clone 4E7, tissue culture supernatant, isotype IgG2a

Synonym(s):

Monoclonal Anti-Mage-11 antibody produced in mouse, Melanoma antigen family A, 11, CT1.11, MAGEA-11, MAGEA11

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

conjugate

unconjugated

antibody form

tissue culture supernatant

antibody product type

primary antibodies

clone

4E7, monoclonal

form

buffered aqueous solution

mol wt

~47 kDa

species reactivity

rat, human, sheep, bovine, monkey, mouse, hamster, canine

technique(s)

immunocytochemistry: suitable
immunoprecipitation (IP): suitable
western blot: 1:250-1:500 using HeLa cell extracts
western blot: suitable

isotype

IgG2a

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

General description

Monoclonal Anti-MAGE-11 (rat IgG2a isotype) is derived from the hybridoma 4E7 produced by the fusion of mouse myeloma cells (P3X63Ag8.653) and splenocytes from rat immunized with a fusion protein corresponding to a fragment of human melanoma associated gene-11 (MAGE-11). In human, the MAGE family of genes includes about 25 genes. The MAGE-A family consists of 12 proteins. This gene is mapped o human chromosome Xq28.

Specificity

Monoclonal Anti-MAGE-11 recognizes human, monkey, bovine, mouse, rat, hamster, sheep, and dog MAGE-11.

Application

Monoclonal Anti- MAGE-11 has been used in:
  • immunoblotting
  • immunoprecipitation
  • immunocytochemistry

Biochem/physiol Actions

Melanoma associated gene-11 (MAGE-A) is identified as one of the cancer-testis antigens in some embryonic tissues. MAGE-A acts as a coregulator of androgen receptor (AR) expression in human female reproductive tract. It is involved in neuronal development and apoptosis. MAGE-A11 has been found mainly in the nucleus of cancer cells. Interestingly, MAGE-11 interferes with tumor angiogenesis and glycolytic switch by affecting the hypoxia-inducible factors (HIFs) activation.

Physical form

Solution in 0.01 M phosphate buffered saline pH 7.4, containing 15 mM sodium azide as a preservative, containing 10% fetal calf serum.

Storage and Stability

Store at –20 °C. For continuous use, the product maybe stored at 2–8 °C for up to one month. For extended storage, freeze at –20 °C in working aliquots. Repeated freezing and thawing, or storage in “frost-free” freezers, is not recommended. If slight turbidity occurs upon prolonged storage, clarify the solution by centrifugation before use. Working dilution samples should be discarded if not used within 12 hours.

Disclaimer

Unless otherwise stated in our catalog, our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class

10 - Combustible liquids

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

The MAGE proteins: emerging roles in cell cycle progression, apoptosis, and neurogenetic disease
Barker PA and Salehi A
Journal of Neuroscience Research, 67(6), 705-712 (2002)
Melanoma antigen-11 inhibits the hypoxia-inducible factor prolyl hydroxylase 2 and activates hypoxic response
Aprelikova O, et al.
Cancer research, 69(2), 616-624 (2009)
Hormone control and expression of androgen receptor coregulator MAGE-11 in human endometrium during the window of receptivity to embryo implantation
Bai S, et al.
Molecular Human Reproduction, 14(2), 107-116 (2007)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service