Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

SAB4200156

Sigma-Aldrich

Anti-SORBS3 antibody, Mouse monoclonal

clone SORB84, purified from hybridoma cell culture

Synonym(s):

Anti-SCAM1, Anti-SH3D4, Anti-VINEXIN

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

mouse

conjugate

unconjugated

antibody form

purified from hybridoma cell culture

antibody product type

primary antibodies

clone

SORB84, monoclonal

form

buffered aqueous solution

mol wt

antigen 37 kDa

species reactivity

monkey, human, rat

concentration

~1.5 mg/mL

technique(s)

western blot: 1-2 μg/mL using HeLa, A549, COS7 or PC12 total cell extracts

UniProt accession no.

shipped in

dry ice

Storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... SORBS3(10174)

General description

Monoclonal Anti-SORBS3 (mouse IgG2b isotype) is derived from the hybridoma SORB84 produced by the fusion of mouse myeloma cells and splenocytes from BALB/c mice immunized with a synthetic peptide. The gene for sorbin and SH3 domain containing 3 (SORBS3) (also known as vinexin) protein is mapped to human chromosome 8p21.3. This protein belongs to the family of adaptor proteins. All the family members share a sorbin homology (SoHo) domain in the N-terminal region and three Src homology 3 (SH3) domains in the C-terminal region. There are three different isoforms of SORBS3 (α, β, and γ) protein that has been characterized. Isoform α and β contain three SH3 domains while the β isoform does not include the N-terminal SoHo domain. The γ isoform of vinexin has been characterized in mouse fetal gonads. It differs from the α isoform in a few amino acids that are missing from the N-terminal region. Vinexin ⓰ is ubiquitously expressed but vinexin α and γ show tissue-specific expression.

Specificity

Monoclonal Anti-SORBS3 recognizes human, monkey, and rat SORBS3.

Application

Anti-SORBS3 antibody, Mouse monoclonal is suitable for immunoblotting.

Biochem/physiol Actions

Vinexin α and β participates in cell signaling and the cytoskeletal structure. The α isoform is known to induce actin stress fiber formation and the β isoform plays a role in cell spreading. Vinexin might enhance the activation of c-Jun N-terminal kinase/stress-activated protein kinase (JNK/SAPK) in response to epidermal growth factor (EGF) stimulation by using its third SH3 domain. The growth-factor-induced signal transductions are controlled by the vinexin-son of sevenless (SOS) interactions. The sorbin homology (SoHo) domain has been shown to bind the lipid raft-associated protein flotillin1. The Src homology 3 (SH3) domain allows the binding of specific signaling and cytoskeletal molecules, including vinculin.

Physical form

Solution in 0.01M phosphate buffered saline pH 7.4, containing 15 mM sodium azide.

Storage and Stability

Store at –20 °C. For continuous use, the product may be stored at 2–8 °C for up to one month. For extended storage, freeze at –20 °C in working aliquots. Repeated freezing and thawing, or storage in “frost-free” freezers, is not recommended. If slight turbidity occurs upon prolonged storage, clarify the solution by centrifugation before use. Working dilution samples should be discarded if not used within 12 hours.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class

10 - Combustible liquids

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Makoto Matsuyama et al.
Genes to cells : devoted to molecular & cellular mechanisms, 10(5), 421-434 (2005-04-20)
Recent loss-of-function and gain-of-function studies have revealed that transcription factor Sox9 is required for testis formation by governing Sertoli cell differentiation, and thereafter regulating transcription of Sertoli marker genes. In the present study, we identified a novel isoform of Vinexin
Noriyuki Kioka et al.
Cell structure and function, 27(1), 1-7 (2002-04-09)
Adaptor proteins, composed of two or more protein-protein interacting modules without enzymatic activity, regulate various cellular functions. Vinexin, CAP/ponsin, and ArgBP2 constitute a novel adaptor protein family. They have a novel conserved region homologous to the active peptide sorbin, as
Samantha E Day et al.
Clinical epigenetics, 8, 77-77 (2016-07-21)
Obesity is a metabolic disease caused by environmental and genetic factors. However, the epigenetic mechanisms of obesity are incompletely understood. The aim of our study was to investigate the role of skeletal muscle DNA methylation in combination with transcriptomic changes

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service