Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

SAB4200154

Sigma-Aldrich

Anti-Importin 9 (C-terminal) antibody produced in rabbit

~1.0 mg/mL, affinity isolated antibody

Synonym(s):

Anti-IMP9, Anti-IPO9, Anti-RAN-binding protein 9, Anti-RANBP9

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

rabbit

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

antigen ~120 kDa

species reactivity

human

concentration

~1.0 mg/mL

technique(s)

immunoprecipitation (IP): 5-10 μg using HEK-293T cell lysates
western blot: 1-2 μg/mL using HeLa cell lysates

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... IPO9(55705)
mouse ... Ipo9(226432)
rat ... Ipo9(304817)

General description

Importin 9 (IPO9) is encoded by the gene mapped to human chromosome 1q32.1. The encoded protein is a molecular transporter and chaperone and is a member of karyopherins super-family. IPO9 has a molecular weight of ~110kDa.
Protein IPO9 exists in two isoforms: a and b in mouse.

Specificity

Anti-Importin 9 (C-terminal) recognizes human Importin 9.

Immunogen

synthetic peptide corresponding to a sequence at the C-terminal part of human Importin 9 conjugated to KLH. The corresponding sequence is identical in mouse and rat.

Application

Anti-Importin 9 (C-terminal) antibody produced in rabbit is suitable for immunoblotting and immunoprecipitation.

Biochem/physiol Actions

Importin 9 (IPO9) promotes transcriptional activity by actively regulating nuclear actin levels. The encoded protein facilitates transport of ribosomal proteins and core histones into the nucleus. Additionally, it acts as an import receptor for c-Jun. Elevated expression of Ipo9 decreased and knockdown of Ipo9 increased basal type I interferon (IFN)-ε expression.
Importins mediate the active transport of cargo molecules through nuclear pore complexes (NPCs). The importins might also protect the cargoes against unnecessary interactions during transit to the nucleus. IPO9 serves as an import factor for protein phosphatase 2A (PP2A) and the hepatocellular carcinoma-associated protein.

Physical form

Solution in 0.01 M phos­phate buffered saline, pH 7.4, containing 15 mM sodium azide.

Storage and Stability

For continuous use, store at 2-8 °C for up to one month. For extended storage, freeze in working aliquots. Repeated freezing and thawing, or storage in “frost-free” freezers, is not recommended. If slight turbidity occurs upon prolonged storage, clarify the solution by centrifugation before use. Working dilutions should be discarded if not used within 12 hours.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class

10 - Combustible liquids

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Yuh Min Chook et al.
Biochimica et biophysica acta, 1813(9), 1593-1606 (2010-10-30)
Proteins in the karyopherin-β family mediate the majority of macromolecular transport between the nucleus and the cytoplasm. Eleven of the 19 known human karyopherin-βs and 10 of the 14S. cerevisiae karyopherin-βs mediate nuclear import through recognition of nuclear localization signals
Novel role for molecular transporter importin 9 in posttranscriptional regulation of IFN-? expression.
Matsumiya T
Journal of Immunology, 191, 1907-1915 (2013)
Stefan Jäkel et al.
The EMBO journal, 21(3), 377-386 (2002-02-02)
Many nuclear transport pathways are mediated by importin beta-related transport receptors. Here, we identify human importin (Imp) 4b as well as mouse Imp4a, Imp9a and Imp9b as novel family members. Imp4a mediates import of the ribosomal protein (rp) S3a, while
Active maintenance of nuclear actin by importin 9 supports transcription.
Dopie J
Proceedings of the National Academy of Sciences of the USA, 109, E544-E552 (2012)
Screening of tumor suppressor genes on 1q31.1-32.1 in Chinese patients with sporadic colorectal cancer.
Zhou CZ
Chinese Medical Journal (English Edition), 121, 2479-2486 (2008)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service