Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

SAB4200145

Sigma-Aldrich

Anti-AMSH-LP/STAMBPL1 antibody produced in rabbit

~1.0 mg/mL, affinity isolated antibody

Synonym(s):

Anti-ALMalpha, Anti-AMSH-FP, Anti-AMSH-like protease, Anti-STAM binding protein-like 1, Anti-associated molecule with the SH3 domain of STAM (AMSH) like protein

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

rabbit

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

~50 kDa

species reactivity

human

packaging

antibody small pack of 25 μL

concentration

~1.0 mg/mL

technique(s)

indirect immunofluorescence: 2-5 μg/mL using human HeLa cells
western blot: 1-2 μg/mL using whole extracts of human PMC2 cells

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

General description

STAM binding protein like 1 (STAMBPL1) or AMSH-LP (AMSH-like protein), is a close homolog of AMSH (Associated Molecule with the SH3 domain of STAM). AMSH and AMSH-LP belong to the Joseph disease protease and JAB1/MPN/Mov34 metalloenzyme (JAMM) domain metalloprotease family of Zn2+-dependent deubiquitinating enzymes (DUBs). STAMBPL1 is mapped to human chromosome 10q23.31.

Specificity

Anti-AMSH-LP/STAMBPL1 recognizes human AMSH-LP/STAMBPL1.

Application

Anti-AMSH-LP/STAMBPL1 antibody produced in rabbit has been used in immunoblotting and immunofluorescence.

Biochem/physiol Actions

STAM binding protein like 1 (STAMBPL1), along with Associated Molecule with the SRC homology 3 domain (SH3) of STAM (AMSH proteins) mediate the deubiquitination of endosomal proteins and specifically cleave lysine-63-linked polyubiquitin chains. AMSH-LP, like AMSH, interacts with clathrin heavy chain and this interaction is essential for its endosomal localization. However, AMSH-LP, unlike AMSH, fails to bind to the SH3 domain of STAM, suggesting that they are not functionally redundant. AMSH-LP positively regulates transforming growth factor (TGF-β) signaling through interaction with inhibitory I-SMADs.

Physical form

Solution in 0.01 M phosphate buffered saline, pH 7.4, containing 15 mM sodium azide

Storage and Stability

Store at –20 °C. For continuous use, store at 2–8 °C for up to one month. For extended storage, freeze in working aliquots. Repeated freezing and thawing, or storage in “frost-free” freezers, is not recommended. If slight turbidity occurs upon prolonged storage, clarify the solution by centrifugation before use. Working dilutions should be discarded if not used within 12 hours.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class

10 - Combustible liquids

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Cloning of a novel signaling molecule, AMSH-2, that potentiates transforming growth factor beta signaling
Ibarrola N, et al.
BMC Cell Biology, 5(1), 2-2 (2004)
Structural basis for specific cleavage of Lys 63-linked polyubiquitin chains
Sato Y, et al.
Nature, 455(7211), 358-362 (2008)
Clathrin anchors deubiquitinating enzymes, AMSH and AMSH-like protein, on early endosomes
Nakamura M, et al.
Genes Cells, 11(6), 593-606 (2006)
Xi Chen et al.
Cancer letters, 456, 49-58 (2019-04-21)
The zinc metalloprotease STAM-binding protein-like 1 (STAMBPL1) has been identified as a deubiquitinase by specifically cleaving Lys-63-linked polyubiquitin chains, but its cellular function remains unclear. Here we described the potential role of STAMBPL1 in suppression of the intrinsic apoptosis. We
Chinyere Ibeawuchi et al.
International journal of molecular sciences, 16(2), 3856-3869 (2015-02-14)
The multifocal nature of prostate cancer (PCa) creates a challenge to patients' outcome prediction and their clinical management. An approach that scrutinizes every cancer focus is needed in order to generate a comprehensive evaluation of the disease, and by correlating

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service