Skip to Content
MilliporeSigma
All Photos(3)

Key Documents

SAB4200139

Sigma-Aldrich

Anti-VILIP-1 (C-terminal) antibody produced in rabbit

enhanced validation

~1.5 mg/mL, affinity isolated antibody

Synonym(s):

Anti-HLP3, Anti-HPCAL3, Anti-HUVISL1, Anti-Hippocalcin-like protein3, Anti-VILIP, Anti-VSNL1, Anti-Visinin-like protein-1 (VLP-1)

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.44

biological source

rabbit

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

antigen ~22 kDa

species reactivity

mouse, human, rat

packaging

antibody small pack of 25 μL

enhanced validation

recombinant expression
Learn more about Antibody Enhanced Validation

concentration

~1.5 mg/mL

technique(s)

immunoprecipitation (IP): 5-10 μg using mouse brain extracts (S1 fraction)
western blot: 1.5-3.0 μg/mL using rat brain extracts (S1 fraction) and HEK-293T cell lysate overexpressing human VILIP-1

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... VSNL1(7447)
mouse ... Vsnl1(26950)
rat ... Vsnl1(24877)

General description

Visinin-like protein-1 (VILIP-1) belongs to a large family of neuronal Ca2+ sensor (NCS) proteins. The VILIP subfamily includes VILIP-1, VILIP-2, VILIP-3, hippocalcin and neurocalcin-δ. VILIP-1 is primarily expressed in the brain, particularly in the hippocampus. NCS proteins possess four EF-hand Ca2+-binding motifs, that facilitate their association with lipid bilayers.

Application

Anti-VILIP-1 (C-terminal) antibody produced in rabbit has been used in immunoblotting and immunoprecipitation.

Biochem/physiol Actions

Visinin-like protein-1 (VILIP-1) associates with the plasma membrane and Golgi membranes and modulates the signaling of cAMP and cGMP in neuronal cells. It upregulates the expression of functional α4β2 neuronal nicotinic acetylcholine receptors in hippocampal neurons. VILIP-1 modulates the activity of guanylyl cyclase B through clathrin dependent receptor cycling, supporting a role for VILIP-1 in membrane trafficking in the central nervous system (CNS). VILIP-1 is implicated in the pathology of CNS disorders including Alzheimer′s disease, schizophrenia and as a biomarker in ischemic stroke. VILIP-1 expression is regulated during induction of hippocampal synaptic plasticity, underlying learning and memory processes. Neuronal Ca2+ sensor (NCS) proteins are involved in a variety of Ca2+-dependent signal transduction processes in neurons.

Physical form

Solution in 0.01 M phos­phate buffered saline, pH 7.4, containing 15 mM sodium azide.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class

10 - Combustible liquids

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Expression of the neuronal calcium sensor visinin-like protein-1 in the rat hippocampus
Zhao C and Braunewell KH
Neuroscience, 153(4), 1202-1212 (2008)
Neuronal Ca2+ sensor protein VILIP-1 affects cGMP signalling of guanylyl cyclase B by regulating clathrin-dependent receptor recycling in hippocampal neurons
Brackmann M, et al.
Journal of Cell Science, 118(11), 2495-2505 (2005)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service