Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

SAB4200138

Sigma-Aldrich

Anti-Synemin (C-terminal region) antibody produced in rabbit

~1.5 mg/mL, affinity isolated antibody

Synonym(s):

Anti-DMN, Anti-Desmuslin, Anti-SYN

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

rabbit

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

~170 kDa

species reactivity

rat

packaging

antibody small pack of 25 μL

concentration

~1.5 mg/mL

technique(s)

western blot: 1.5-3.0 μg/mL using using rat bladder extract (S1 fraction).

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

General description

Synemin exists in two isoforms, α and β.
Synemin is a 230kDa intermediate filament protein (IFp). It is expressed along with desmin and vimentin filaments in muscle cells. In mouse, synemin is encoded by the gene mapped in B5 region of chromosome 7. Alternative splicing of this gene produces three different isoforms with different molecular mass namely, high (180kDa), medium (150kDa) and low (41kDa). High and medium isoforms are expressed in embryonic mouse, whereas, low molecular weight protein is expressed in adult mouse. Synemin comprises ∼310-amino acid rod domain characteristic of IF proteins and 1,290 amino acid tail domain at its C- terminal.

Specificity

Anti-Synemin (C-terminal region) specifically recognizes rat synemin.

Application

Anti-Synemin (C-terminal region) antibody produced in rabbit is suitable for immunoblotting.

Biochem/physiol Actions

α-Synemin is essential for the stabilization of junctional complexes at sarcolemma, in neonates. β-Synemin controls the connection of desmin with Z-disks. Synemin is also involved in protein phosphorylation, for regulating key signaling cascades in the heart. Mild myopathy has been observed in synemin knockout mice.
Synemin in association with either desmin or vimentin plays a vital role in cytoskeletal organization and also facilitates function of intermediate filaments in erythrocytes and muscle. Interaction of synemin and vinculin aids in attachment of intermediate filaments (Ifs) to the cell membrane.

Physical form

Solution in 0.01 M phosphate buffered saline, pH 7.4, containing 15 mM sodium azide.

Storage and Stability

Store at -20 °C.For continuous use, store at 2-8 °C for up to one month. For extended storage, freeze in working aliquots. Repeated freezing and thawing, or storage in “frost-free” freezers,is not recommended. If slight turbidity occurs upon prolonged storage, clarify the solution by centrifugation.Working dilutions should be discarded if not used within 12 hours.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class

10 - Combustible liquids

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Synemin: a new high molecular weight protein associated with desmin and vimentin filaments in muscle
Granger BL and Lazarides E
Cell, 22, 727-738 (1980)
Synemin and vimentin are components of intermediate filaments in avian erythrocytes.
Granger BL
The Journal of Cell Biology, 92, 299-312 (1982)
Karla P García-Pelagio et al.
Journal of molecular and cellular cardiology, 114, 354-363 (2017-12-17)
Cardiomyopathies have been linked to changes in structural proteins, including intermediate filament (IF) proteins located in the cytoskeleton. IFs associate with the contractile machinery and costameres of striated muscle and with intercalated disks in the heart. Synemin is a large
Synemin May Function to Directly Link Muscle Cell Intermediate Filaments to Both Myofibrillar Z-lines and Costameres*
Bellin RM
The Journal of Biological Chemistry, 276, 32330-32337 (2001)
The mouse synemin gene encodes three intermediate filament proteins generated by alternative exon usage and different open reading frames
Xue ZG
Experimental Cell Research, 298, 431-444 (2004)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service