Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

SAB4200134

Sigma-Aldrich

Anti-PNK antibody ,Mouse monoclonal

clone H101.5, purified from hybridoma cell culture

Synonym(s):

Anti-Bifunctional polynucleotide phosphatase/kinase, Anti-DNA 5′-kinase/3′-phosphatase, Anti-PNKP, Anti-Polynucleotide kinase 3′-phosphatase

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.44

biological source

mouse

conjugate

unconjugated

antibody form

purified from hybridoma cell culture

antibody product type

primary antibodies

clone

H101.5, monoclonal

form

buffered aqueous solution

mol wt

antigen ~55 kDa

species reactivity

monkey, human, bovine

packaging

antibody small pack of 25 μL

concentration

~1.0 mg/mL

technique(s)

western blot: 2.5-5.0 μg/mL using whole extracts of A549 cells

isotype

IgG1

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... PNKP(11284)

General description

Monoclonal Anti-PNK (mouse IgG1 isotype) is derived from the hybridoma H101.5 produced by the fusion of mouse myeloma cells and splenocytes from BALB/c mice immunized with a human polynucleotide kinase 3′-phosphatase (PNK) recombinant protein. PNK comprises of a central phosphatase domain that is flanked by an N-terminal forkhead-associated (FHA) domain and a C-terminal kinase domain on either sides.

Immunogen

human PNK recombinant protein.

Application

Monoclonal Anti-PNK antibody produced in mouse has been used in immunoblotting and immunoprecipitation.

Biochem/physiol Actions

PNK is a mammalian DNA repair enzyme which catalyzes the phosphorylation of DNA 5′-hydroxyl termini and the dephosphorylation of 3′-phosphate termini. PNK is involved in the single-strand break repair pathway and the non-homologous end joining pathway for double strand break repair. Mutations in PNK result in neurological abnormalities.
Polynucleotide kinase 3′-phosphatase (PNKP or PNK) is a nuclear protein that repairs DNA strand breaks induced by oxidative damage or ionizing radiation. Genetic alterations in PNKP have been linked to seizures, microcephaly and deficiencies in DNA repair mechanisms .

Physical form

Solution in 0.01 M phosphate buffered saline, pH 7.4, containing 15 mM sodium azide.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class

10 - Combustible liquids

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Production, characterization, and epitope mapping of monoclonal antibodies against human polydeoxyribonucleotide kinase
Fanta M, et al.
Hybridoma, 20(4), 237-242 (2001)
PNKP knockdown by RNA interference inhibits herpes simplex virus-1 replication in astrocytes
Yue L, et al.
Virologica Sinica, 28(6), 345-351 (2013)
Human polynucleotide kinase participates in repair of DNA double-strand breaks by nonhomologous end joining but not homologous recombination
Karimi BF, et al.
Cancer research, 67(14), 6619-6625 (2007)
N K Bernstein et al.
Anti-cancer agents in medicinal chemistry, 8(4), 358-367 (2008-05-14)
The cytotoxicity of many antineoplastic agents is due to their capacity to damage DNA and there is evidence indicating that DNA repair contributes to the cellular resistance to such agents. DNA strand breaks constitute a significant proportion of the lesions
A Jilani et al.
The Journal of biological chemistry, 274(34), 24176-24186 (1999-08-14)
Mammalian polynucleotide kinases catalyze the 5'-phosphorylation of nucleic acids and can have associated 3'-phosphatase activity, predictive of an important function in DNA repair following ionizing radiation or oxidative damage. The sequences of three tryptic peptides from a bovine 60-kDa polypeptide

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service