Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

SAB4200131

Sigma-Aldrich

Monoclonal Anti-PSMD1 antibody produced in mouse

~1.0 mg/mL, clone PSMD1-16, purified immunoglobulin

Synonym(s):

Anti-Rpn2, Anti-S1, Anti-p112, Anti-proteasome (prosome, macropain) 26S non-ATPase subunit 1

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

mouse

conjugate

unconjugated

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

PSMD1-16, monoclonal

form

buffered aqueous solution

mol wt

~105 kDa

species reactivity

mouse, canine, human, rat

concentration

~1.0 mg/mL

technique(s)

immunoprecipitation (IP): suitable
western blot: 1-2 μg/mL using using whole extracts of human HEK-293T cells.

isotype

IgG1

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... PSMD1(5707)
mouse ... Psmd1(70247)
rat ... Psmd1(83806)

Physical form

Solution in 0.01 M phosphate buffered saline, pH 7.4, containing 15 mM sodium azide.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class

10 - Combustible liquids

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service