Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

SAB4200121

Sigma-Aldrich

Anti-Annexin 4 antibody,Mouse monoclonal

clone ANX47, purified from hybridoma cell culture

Synonym(s):

Anti-ANX4, Anti-ANXA4, Anti-PIG28, Anti-Placental anticoagulant protein II, Anti-ZAP36, Anti-Zymogen granule membrane-associated protein, 36-KD

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203

biological source

mouse

conjugate

unconjugated

antibody form

purified from hybridoma cell culture

antibody product type

primary antibodies

clone

ANX47, monoclonal

form

buffered aqueous solution

mol wt

antigen 34 kDa

species reactivity

mouse, bovine, human, monkey

packaging

antibody small pack of 25 μL

concentration

~1.5 mg/mL

technique(s)

immunohistochemistry: suitable
western blot: 2-4 μg/mL using A431 total cell extracts

isotype

IgG1

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... ANXA4(307)

General description

Annexin A4 is encoded by the gene mapped to human chromosome 2p13.3. The encoded protein belongs to the Ca2+-regulated and phospholipid-binding annexin superfamily proteins. Annexin A4 is highly expressed in epithelial cells.
Monoclonal Anti-Annexin 4 (mouse IgG1 isotype) is derived from the hybridoma ANX47 produced by the fusion of mouse myeloma cells and splenocytes from BALB/c mice immunized with a recombinant human Annexin 4 protein.

Immunogen

recombinant human Annexin 4 protein

Application

The product may be used in several immunochemical techniques including immunoblotting and immunohistochemistry.

Biochem/physiol Actions

Elevated expression of annexin A4 leads to development of various cancers such as colorectal cancer (CRC), esophageal squamous cell carcinoma (ESCC), endometrial carcinoma (EC), gastric cancer (GC), chemoresistant lung cancer (LC), malignant mesothelioma (MM), renal cell carcinoma (RCC), ovarian clear cell carcinoma (OCCC), cholangiocarcinoma, hepatocellular carcinoma (HCC), breast cancer (BC) and laryngeal cancer. Thus, annexin A4 can be used in diagnosis, prognosis and treatment of these cancers. In addition, the encoded protein acts as a tumor suppressor for prostate cancer (PCa) and has a contradictory role for pancreatic cancer (PCC).
The major involvement of Annexin 4 is seen in membrane permeability, exocytosis and regulation of ion channels. Its role in membrane fluidity and membrane trafficking may explain in part the involvement of Annexin 4 in modulating drug resistance in cancer cells. Indeed, Annexin 4 was found to confer chemoresistance in ovarian cancer cells in part by enhancing drug efflux.

Physical form

Solution in 0.01M phosphate buffered saline pH 7.4, containing 15 mM sodium azide.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class

10 - Combustible liquids

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Enhanced expression of Annexin A4 in clear cell carcinoma of the ovary and its association with chemoresistance to carboplatin
Kim A, et al.
International Journal of Cancer. Journal International Du Cancer, 125(10), 2316-2322 (2009)
Proteome of the early embryo-maternal dialogue in the cattle uterus
Munoz M, et al.
Journal of Proteome Research, 11(2), 751-766 (2011)
Fhit Delocalizes Annexin A4 from Plasma Membrane to Cytosol and Sensitizes Lung Cancer Cells to Paclitaxel
Gaudio E
PLoS ONE (2013)
Annexin A4 and cancer.
Wei B
Clinica Chimica Acta; International Journal of Clinical Chemistry, 447, 72-78 (2015)
Gene Expression and Proteomic Analysis of Pancreatic Cancer: a Recent Update
CHEN R U
Cancer, Genomics, and Proteomics, 1-10 (2006)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service