Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

SAB4200116

Sigma-Aldrich

Anti-Coronin-1C (C-terminal) antibody produced in rabbit

~1.5 mg/mL, affinity isolated antibody

Synonym(s):

Anti-CORO1C, Anti-Coronin-3, Anti-HCRNN4

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

rabbit

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

antigen ~55 kDa

species reactivity

human, mouse

concentration

~1.5 mg/mL

technique(s)

indirect immunofluorescence: 5-10 μg/mL using NIH3T3 cells
western blot: 1.5-3.0 μg/mL using A431 cell extract

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... CORO1C(23603)

General description

Coronin 1C (CORO1C) is encoded by the gene mapped to human chromosome 12q24.1. The encoded protein belongs to coronin-like family of proteins. CORO1C consists of 474 amino acids and is ubiquitously expressed in human tissues. CORO1C contains conserved five WD repeats in its N-terminal end and a coiled-coil motif in its C-terminal end.The protein is mainly present at the sites of active actin dynamics including lamellipodia and membrane ruffles.

Biochem/physiol Actions

Coronin 1C (CORO1C) plays a vital role in cytokinesis, motility and signal transduction. It is also implicated in regulation of various cellular processes such as cell proliferation, migration, invadopodia formation and invasion in glioblastoma cells. CORO1C acts as a predictive biomarker for hepatocellular carcinoma (HCC) invasive behavior. The encoded protein negatively regulates cell-matrix adhesion and intestinal epithelial cells (IECs) migration via FAK-mediated signaling.

Physical form

Solution in 0.01 M phos­phate buffered saline, pH 7.4, containing 15 mM sodium azide.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class

10 - Combustible liquids

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Coronin-1C is a novel biomarker for hepatocellular carcinoma invasive progression identified by proteomics analysis and clinical validation.
Wu L
Journal of Experimental & Clinical Cancer Research (2010)
Coronin 1C negatively regulates cell-matrix adhesion and motility of intestinal epithelial cells.
Samarin SN
Biochemical and Biophysical Research Communications, 391, 394-400 (2010)
Isolation and chromosomal assignment of a novel human gene, CORO1C, homologous to coronin-like actin-binding proteins.
Iizaka M
Cytogenetics and Cell Genetics, 88, 221-224 (2000)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service