Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

SAB4200111

Sigma-Aldrich

Anti-TARBP2 antibody produced in rabbit

~1.0 mg/mL, affinity isolated antibody, buffered aqueous solution

Synonym(s):

Anti-TRBP, Anti-TRBP1, Anti-Tar (HIV-1) RNA binding protein 2

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352203
NACRES:
NA.41

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

antigen ~30 kDa

species reactivity

human

packaging

antibody small pack of 25 μL

concentration

~1.0 mg/mL

technique(s)

indirect immunofluorescence: 2.5-5 μg/mL using paraformaldehyde fixed HeLa cells
western blot: 2-4 μg/mL using lysates of HEK-293T cells

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... TARBP2(6895)
mouse ... 21357(21357)
rat ... Tarbp2(363006)

General description

The transactivating response (TAR) RNA-binding protein (TRBP) (also known as TARBP2) gene is mapped to human chromosome 12q13.13. The encoded protein exists in two isoforms, TRBP1 and TRBP2, where TRBP2 has 21 additional amino acids. It is composed of two double stranded RNA binding domains (dsRBDs), a KR-helix motif within dsRBD2 and a Medipal domain that binds Moesin-ezrin-radixin-like protein (Merlin), Dicer and protein activator of the interferon-induced protein kinase (PACT). TRBP is a component of the RNA-induced silencing complex (RISC) loading complex (RLC), also known as the micro-RNA (miRNA) loading complex (miRLC), composed of Dicer and argonaute (Ago).

Specificity

Anti-TRBP2 recognizes human TRBP2.

Immunogen

synthetic peptide corresponding to amino acids 183-195 of human TRBP2 conjugated to KLH. The corresponding sequence is identical in mouse and rat.

Application

Anti-TARBP2 has been used in immunoblotting and might also be used in immunofluorescence.

Biochem/physiol Actions

The transactivating response (TAR) RNA-binding protein (TRBP) (also known as TARBP2) binds and activates the human immunodeficiency virus (HIV)-1 with TAR RNA. It plays a role in spermatogenesis, cell growth, oncogenesis and viral replication. Targeted disruption of the TARBP2 gene, causes growth defects, sterility problems and severe oligospermic effects in mice.

Physical form

Solution in 0.01 M phos­phate buffered saline, pH 7.4, containing 15 mM sodium azide.

Storage and Stability

Store at –20 °C. For continuous use, the product may be stored at 2–8 °C for up to one month. For extended storage, freeze in working aliquots at –20 °C. Repeated freezing and thawing is not recommended. Storage in “frost-free” freezers is also not recommended. If slight turbidity occurs upon prolonged storage, clarify the solution by centrifugation before use. Working dilutions should be discarded if not used within 12 hours.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class

10 - Combustible liquids


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Organization of the human tarbp2 gene reveals two promoters that are repressed in an astrocytic cell line
Bannwarth S, et al.
The Journal of Biological Chemistry, 276(52), 48803-48813 (2001)
Up-regulation and worse prognostic marker of cytoplasmic TARBP2 expression in obstinate breast cancer
Lin X, et al.
Medical Oncology (Northwood, London, England), 31(4), 868-868 (2014)
Merlin, a tumor suppressor, interacts with transactivation-responsive RNA-binding protein and inhibits its oncogenic activity
Lee JY, et al.
The Journal of biological chemistry, 279(29), 30265-30273 (2004)
Characterization of the TRBP domain required for dicer interaction and function in RNA interference
Daniels SM, et al.
BMC Molecular Biology, 10(1), 1-13 (2009)
In vitro reconstitution of the human RISC-loading complex
MacRae IJ, et al.
Proceedings of the National Academy of Sciences of the USA, 105(2), 512-517 (2008)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service