Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

SAB4200110

Sigma-Aldrich

Anti-PIWIL1 (internal region) antibody produced in rabbit

~1.0 mg/mL, buffered aqueous solution

Synonym(s):

Anti-HIWI, Anti-MIWI, Anti-PIWI-like 1

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

rabbit

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

~95 kDa

species reactivity

human, rat, mouse

concentration

~1.0 mg/mL

technique(s)

immunohistochemistry: suitable using heat-retrieved formalin-fixed, paraffin-embedded human colon carcinoma sections and biotin/ExtrAvidin®-Peroxidase staining system
western blot: 1-2 μg/mL using rat and mouse testis extracts

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... PIWIL1(9271)
mouse ... Piwil1(57749)
rat ... Piwil1(363912)

General description

The PIWIL1 (Piwi like RNA-mediated gene silencing 1) gene encodes an 861 amino acid protein, which belongs to the Piwi subfamily of Argonaute proteins. This gene is mapped to human chromosome 12q24.33. The Piwi proteins are mostly localized to the germline, where they bind to PIWI-interacting RNAs (piRNAs). The Piwi proteins contain the Piwi-Argonaute-Zwille (PAZ) domain and the PIWI domain.

Specificity

Anti-PIWIL1 (internal region) recognizes human, mouse, and rat PIWIL1.

Application

Anti-PIWIL1 (internal region) antibody produced in rabbit is suitable for immunoblotting and immunohistochemistry.

Biochem/physiol Actions

The Piwi proteins are important for the biogenesis and function of small non-coding RNAs. These proteins and PIWI-interacting RNAs (piRNAs) have been implicated in epigenetic control of gene expression, transposon silencing, gene expression, and translation regulation. The Piwi proteins significantly participate in germline development and gametogenesis of various metazoan species. These proteins undergo symmetrical dimethyl arginines (sDMAs) modification by protein methyltransferase 5 (PRMT5).

Physical form

Solution in 0.01 M phosphate buffered saline, pH 7.4, containing 15 mM sodium azide

Storage and Stability

Store at –20 °C. For continuous use, store at 2–8 °C for up to one month. For extended storage, freeze in working aliquots. Repeated freezing and thawing, or storage in “frost-free” freezers, is not recommended. If slight turbidity occurs upon prolonged storage, clarify the solution by centrifugation. Working dilutions should be discarded if not used within 12 hours.

Legal Information

ExtrAvidin is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class

10 - Combustible liquids

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

The Biogenesis and Function PIWI Proteins and piRNAs: Progress and Prospect
Thomson T and Lin H
Annual Review of Cell and Developmental Biology, , number=(25), 355?376-355?376 (2009)
Arginine methylation of Piwi proteins catalysed by dPRMT5 is required for Ago3 and Aub stability
Kirino Y, et al.
Nature Cell Biology, 11(5) (2009)
Expression status of PIWIL1 as a prognostic marker of colorectal cancer
Sun R, et al.
Disease Markers, 2017(5) (2017)
The Argonaute protein family
Hock J and Gunter M
Genome Biology, 9(2), 210-210 (2008)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service