Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

SAB4200109

Sigma-Aldrich

Anti-PIWIL1 antibody produced in rabbit

~1.0 mg/mL, affinity isolated antibody

Synonym(s):

Anti-HIWI, Anti-MIWI, Anti-PIWI-like 1

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

rabbit

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

antigen ~95 kDa

species reactivity

human, mouse

packaging

antibody small pack of 25 μL

concentration

~1.0 mg/mL

technique(s)

immunohistochemistry: 10-20 μg/mL using heat-retrieved formalin-fixed, paraffin-embedded mouse testis and biotin / ExtrAvidinTM-Peroxidase staining system.
western blot: 1-2 μg/mL using lysates of HEK-293Tcells over expressing human PIWIL1.

UniProt accession no.

shipped in

dry ice

Storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... PIWIL1(9271)

General description

The PIWI-like protein 1 (PIWIL1) belongs to the Piwi subfamily.gene is mapped to human chromosome 12q24.33. It is expressed in the cytosol of spermatocytes and spermatids. PIWI-like protein 1 (PIWIL1) is also an Argonaute protein family member.

Specificity

Anti-PIWIL1 recognizes human and mouse PIWIL1.

Application

Anti-PIWIL1 antibody produced in rabbit has been used in western blotting and immunohistochemistry.

Biochem/physiol Actions

The PIWI-like protein 1 (PIWIL1) mediates spermatogenesis. Its aberrant expression favors tumor progression inadenocarcinoma. PIWI-like protein 1 (PIWIL1) plays a key role in embryogenesis, differentiation, stem cell proliferation, growth, and development. High levels of PIWIL1 expression is implicated in the colorectal cancer invasion.

Physical form

Solution in 0.01 M phos­phate buffered saline, pH 7.4, containing 15 mM sodium azide.

Storage and Stability

Store at –20 °C. For continuous use, the product may be stored at 2–8 °C for up to one month. For extended storage, freeze at –20 °C in working aliquots. Repeated freezing and thawing, or storage in “frost-free” freezers, is not recommended. If slight turbidity occurs upon prolonged storage, clarify the solution by centrifugation before use. Working dilutions should be discarded if not used within 12 hours.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class

12 - Non Combustible Liquids

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Expression of hiwi gene in human gastric cancer was associated with proliferation of cancer cells
Liu X, et al.
International Journal of Cancer. Journal International Du Cancer, 118(8), 1922-1929 (2006)
Expression status of PIWIL1 as a prognostic marker of colorectal cancer
Sun R, et al.
Disease Markers, 2017(1) (2017)
Cancer-testis gene PIWIL 1 promotes cell proliferation, migration, and invasion in lung adenocarcinoma
Xie K, et al.
Cancer medicine, 7(1), 157-166 (2018)
Circ_101064 regulates the proliferation, invasion and migration of glioma cells through miR-154-5p/PIWIL1 axis
Zhou H, et al.
Biochemical and biophysical research communications, 523(3), 608-614 (2020)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service