Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

SAB4200099

Sigma-Aldrich

Anti-Gapex-5 (N-terminal) antibody produced in rabbit

~1.0 mg/mL, affinity isolated antibody

Synonym(s):

Anti-GAPVD1, Anti-GTPase-activating protein and VPS9 domains 1, Anti-RAB5 exchange factor, Anti-RAP6 (rab5-activating protein 6)

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

rabbit

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

antigen ~180 kDa

packaging

antibody small pack of 25 μL

concentration

~1.0 mg/mL

technique(s)

western blot: 1-2 μg/mL using whole extracts of human HeLa cells

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

General description

The GTPase activating protein and VPS9 domains 1 (Gapex-5) protein, originally known as receptor-mediated endocytosis protein 6 (RME-6)/Rab5-activating protein 6 (RAP6), is a guanine nucleotide exchange factor (GEF) for the Rab5 subfamily of guanine triphosphatase (GTPases). Gapex-5 is an evolutionarily conserved protein with Rab5 GEF domain, the N-terminal Ras GTP hydrolysis-activating protein (GAP) domain and a C-terminal Vps9 domain (a Rab5 guanine diphosphate (GDP)/GTP exchange factor). Gapex-5 is localized on the plasma membrane and on endosomes. This gene is mapped to human chromosome 9q33.3.

Specificity

Anti-Gapex-5 (N-terminal) recognizes human Gapex-5.

Application

Anti-Gapex-5 (N-terminal) antibody produced in rabbit is suitable for immunoblotting.

Biochem/physiol Actions

The GTPase activating protein and VPS9 domains 1 (Gapex-5) protein has guanine nucleotide exchange factor (GEF) activity specific for Rab5 and guanine triphosphate (GTP) hydrolysis-activating protein (GAP) activity specific for Ras. It is involved in multiple processes including receptor-mediated endocytosis, insulin receptor internalization, glucose transporter 4 (Glut4) trafficking in adipocytes, epidermal growth factor receptor (EGFR) trafficking and degradation and phagosome maturation.

Physical form

Solution in 0.01 M phos­phate buffered saline, pH 7.4, containing 15 mM sodium azide.

Storage and Stability

Store at –20 °C. For continuous use, the product may be stored at 2–8 °C for up to one month. For extended storage, freeze in working aliquots at –20 °C. Repeated freezing and thawing is not recommended. Storage in “frost-free” freezers is also not recommended. If slight turbidity occurs upon prolonged storage, clarify the solution by centrifugation before use. Working dilutions should be discarded if not used within 12 hours.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class

10 - Combustible liquids

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Gapex-5, a Rab31 guanine nucleotide exchange factor that regulates Glut4 trafficking in adipocytes
Lodhi IJ, et al.
Cell Metabolism, 5(1) (2007)
Imaging of Rab5 activity identifies essential regulators for phagosome maturation
Kitano M, et al.
Nature, 453(7192), 241-245 (2008)
GAPex-5 mediates ubiquitination, trafficking, and degradation of epidermal growth factor receptor
Su X, et al.
The Journal of biological chemistry, 282(29), 21278-21284 (2007)
Rab5-activating protein 6, a novel endosomal protein with a role in endocytosis
Hunker CM, et al.
Biochemical and biophysical research communications, 340(3), 967-975 (2006)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service