Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

SAB4200098

Sigma-Aldrich

Anti-Gapex-5 antibody produced in rabbit

~1.0 mg/mL, affinity isolated antibody

Synonym(s):

Anti-GAPVD1, Anti-GTPase-activating protein and VPS9 domains 1, Anti-RAB5 exchange factor, Anti-RAP6 (rab5-activating protein 6)

Sign Into View Organizational & Contract Pricing

Select a Size

200 μL
$422.00

$422.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
200 μL
$422.00

About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

$422.00


Check Cart for Availability

Request a Bulk Order

biological source

rabbit

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

antigen ~180 kDa

species reactivity

human

packaging

antibody small pack of 25 μL

concentration

~1.0 mg/mL

technique(s)

immunoprecipitation (IP): 5-10 μg using lysates of human HEK-293T cells
western blot: 1-2 μg/mL using whole extracts of human HeLa cells

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... GAPVD1(26130)
mouse ... Gapvd1(66691)

General description

GTPase-activating protein and VPS9 domain-containing protein 1 (GAPVD1) also called Rab5-activating protein 6 (RAP6) or Gapex-5 gene is mapped to the human chromosome 9q33.3. It has a carboxy-terminal Vps9-like domain and an amino-terminal Ras GTPase-accelerating proteins (GAP)-like domain. GAPVD1 is present in the plasma membrane and endosomes.

Specificity

Anti-Gapex-5 recognizes human Gapex-5.

Application

Anti-Gapex-5 antibody produced in rabbit may be used in immunoblotting and immunoprecipitation.

Biochem/physiol Actions

Rab5-activating protein 6 (RAP6) or Gapex-5 is an evolutionary conserved protein and its C-terminal Vps9 domain is responsible for the interaction with guanosine diphosphate GDP bound Rab5 and an N-terminal Ras GTPase-accelerating proteins (GAP) domain. Gapex-5 has guanine nucleotide exchange factor (GEF) activity specific for Rab5 and GAP activity specific for Ras. It is involved in multiple processes including receptor-mediated endocytosis, insulin receptor internalization and glucose transporter type 4 (Glut4) trafficking in adipocytes. RAP6 is also essential for epidermal growth factor receptor (EGFR) trafficking and degradation and phagosome maturation.

Physical form

Solution in 0.01 M phos­phate buffered saline, pH 7.4, containing 15 mM sodium azide.

Storage and Stability

Store at –20 °C. For continuous use, the product may be stored at 2–8 °C for up to one month. For extended storage, freeze in working aliquots at –20 °C. Storage in “frost-free” freezers is also not recommended. If slight turbidity occurs upon prolonged storage, clarify the solution by centrifugation before use. Working dilutions should be discarded if not used within 12 hours.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class

10 - Combustible liquids

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Insulin stimulates phosphatidylinositol 3-phosphate production via the activation of Rab5
Lodhi IJ, et al.
Molecular Biology of the Cell, 19(7), 2718-2728 (2008)
GAPex-5 mediates ubiquitination, trafficking, and degradation of epidermal growth factor receptor
Su X, et al.
The Journal of biological chemistry, 282(29), 21278-21284 (2007)
Imaging of Rab5 activity identifies essential regulators for phagosome maturation
Kitano M, et al.
Nature, 453(7192), 241-245 (2008)
Rab5-activating protein 6, a novel endosomal protein with a role in endocytosis
Hunker CM, et al.
Biochemical and biophysical research communications, 340(3), 967-975 (2006)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service