Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

SAB4200088

Sigma-Aldrich

Anti-DGCR8 (C-terminal) antibody produced in rabbit

~1.0 mg/mL, affinity isolated antibody

Synonym(s):

Anti-DGCRK8, Anti-DiGeorge syndrom critical region 8

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

rabbit

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

antigen ~100 kDa

species reactivity

human

packaging

antibody small pack of 25 μL

concentration

~1.0 mg/mL

technique(s)

immunoprecipitation (IP): 2.5-5 μg using lysates of HEK-293T cells over expressing human DGCR8
indirect immunofluorescence: 2-5 μg/mL using paraformaldehyde fixed HEK-293T cells over expressing human DGCR8
western blot: 1-2 μg/mL using lysates of HEK-293T cells over expressing human DGCR8

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... DGCR8(54487)
mouse ... Dgcr8(94223)

General description

DGCR8 (DGCR8 microprocessor complex subunit) is a double-stranded RNA-binding protein mapped to the chromosome 22q11.2. It is composed of a WW domain and two double-stranded RNA-binding domains (dsRBDs).[1]

Application

Anti-DGCR8 (C-terminal) antibody produced in rabbit has been used in western blot assay to study the subcellular localization of KSRP (KH-type splicing regulatory protein) in RACK1 (receptor for activated protein kinase C)-knockdown cells. It has been used in Western blotting to study the phosphorylation status of TRBP (TAR RNA binding protein) in a study of glucose tolerance through inhibition of microRNA maturation in miRNA103 transgenic mice.[2] Anti-DGCR8 (C-terminal) antibody produced in rabbit is also suitable for immunoprecipitation at a concentration of 2.5-5μg using lysates of HEK-293T cells over expressing human DGCR8, indirect immunofluorescence at the concentration of 2-5μg/mL using paraformaldehyde fixed HEK-293T cells over expressing human DGCR8 and western blot at a dilution of 1-2μg/mL using lysates of HEK-293T cells over expressing human DGCR8.
Applications in which this antibody has been used successfully, and the associated peer-reviewed papers, are given below.
Western Blotting (1 paper)

Biochem/physiol Actions

DGCR8 (DGCR8 microprocessor complex subunit) participates in the biogenesis of microRNA (miRNA, miR) as a major component of the microprocessor complex. DGCR8 provides guidance to the RNase III enzyme, Drosha during rRNA processing. It forms the microprocessor complex by binding to the RNase III enzyme Drosha, which further converts long primary miRNAs (pri-miRNAs) into short hairpins called precursor miRNAs (pre-miRNAs). The processed hairpins finally are transported to the cytoplasm for further processing by Dicer into mature miRNAs. DGCR8 is also required for global gene regulation and silencing of embryonic stem cell self-renewal. Deletion of DGCR8 chromosomal location has been reported in the DiGeorge and velocardiofacial syndrome.[1]

Physical form

Solution in 0.01 M phos­phate buffered saline, pH 7.4, containing 15 mM sodium azide.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class

10 - Combustible liquids

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Motoko Ohno et al.
Scientific reports, 3, 2553-2553 (2013-08-31)
Polyphenols are representative bioactive substances with diverse biological effects. Here, we show that apigenin, a flavonoid, has suppressive effects on microRNA (miRNA) function. The effects were mediated by impaired maturation of a subset of miRNAs, probably through inhibition of the
Yangming Wang et al.
Nature genetics, 39(3), 380-385 (2007-01-30)
The molecular controls that govern the differentiation of embryonic stem (ES) cells remain poorly understood. DGCR8 is an RNA-binding protein that assists the RNase III enzyme Drosha in the processing of microRNAs (miRNAs), a subclass of small RNAs. Here we
Motoyuki Otsuka et al.
PloS one, 6(9), e24359-e24359 (2011-09-22)
MicroRNAs (miRNAs) are important regulators of gene expression that control physiological and pathological processes. A global reduction in miRNA abundance and function is a general trait of human cancers, playing a causal role in the transformed phenotype. Here, we sought
Chantal Sellier et al.
PloS one, 9(8), e103884-e103884 (2014-08-02)
Deletion of the 1.5-3 Mb region of chromosome 22 at locus 11.2 gives rise to the chromosome 22q11.2 deletion syndrome (22q11DS), also known as DiGeorge and Velocardiofacial Syndromes. It is the most common micro-deletion disorder in humans and one of

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service