Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

SAB4200083

Sigma-Aldrich

Monoclonal Anti-PAX9 antibody produced in rat

~1.0 mg/mL, clone PAX9 7C2, purified immunoglobulin

Synonym(s):

Anti-Paired box 9, Anti-STHAG3

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

rat

conjugate

unconjugated

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

PAX9 7C2, monoclonal

form

buffered aqueous solution

mol wt

antigen ~36 kDa

species reactivity

mouse, human

packaging

antibody small pack of 25 μL

concentration

~1.0 mg/mL

technique(s)

immunohistochemistry: suitable
western blot: 0.25-0.5 μg/mL using H1299 cell extracts

isotype

IgG1

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

mouse ... Pax9(18511)

General description

Monoclonal Anti-PAX9 (rat IgG1 isotype) is derived from the hybridoma paired box 9 (PAX9) 7C2 produced by the fusion of mouse myeloma cells (P3X63Ag8.653) and splenocytes from rat immunized with a recombinant fragment corresponding to a fragment of mouse PAX9 coupled to myelin basic protein (MBP). The PAX gene family encodes a group of transcription factors containing a 128-aa paired DNA-binding domain, and in some cases, a paired-type homeodomain. Nine PAX genes (PAX1-PAX9) has been observed in mammals. Pax9 and Pax1 show high similarities. Pax9 is found in all vertebrates including chick, zebrafish, mouse and man.

Application

Monoclonal Anti-PAX9 antibody produced in rat has been used immunoblotting and immunohistochemistry.

Biochem/physiol Actions

Paired box 9 (PAX9) protein is known to play a role in the formation of various organs and skeletal elements. Mutations in either the PAX1 or the PAX9 genes may produce an inherited skeletal disorder such as the Jarcho-Levin syndrome. Interestingly, progressive loss of PAX9 expression was correlated with increasing malignancy of dysplastic and cancerous epithelium of human esophagus and lung cancer.

Target description

Anti-PAX9, paired box 9, is a member of the paired box (PAX) family of transcription factors. Members of this gene family typically contain a paired box domain, an octapeptide, and a paired-type homeodomain. These genes play critical roles during fetal de

Physical form

Solution in 0.01M phosphate buffered saline pH 7.4, containing 15 mM sodium azide.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class

10 - Combustible liquids

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Pax1 and Pax9 activate Bapx1 to induce chondrogenic differentiation in the sclerotome
Rodrigo I, et al.
Development, 130(3), 473?482-473?482 (2003)
Aberrant Pax1 and Pax9 expression in Jarcho-Levin syndrome: Report of two Caucasian siblings and literature review
Bannykh SI, et al.
American Journal of Medical Genetics. Part A, 120(2), 241-246 (2003)
Bingxu Tan et al.
Molecular medicine reports, 16(1), 806-816 (2017-06-01)
Abnormal paired box 9 (PAX9) expression is associated with tumorigenesis, cancer development, invasion and metastasis. The present study investigated the prognostic significance of PAX9 in esophageal squamous cell carcinoma (ESCC) and its role in predicting radiation sensitivity. A total of
Getting your Pax straight: Pax proteins in development and disease
Chi N and Epstein JA
Trends in Genetics, 18(1), 41-47 (2002)
Prognostic value of PAX9 in patients with esophageal squamous cell carcinoma and its prediction value to radiation sensitivity
Tan B, et al.
Molecular Medicine Reports, 16(1), 806-816 (2017)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service