Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

SAB4200077

Sigma-Aldrich

Anti-Coronin-1C (C-terminal)

~1.5 mg/mL, affinity isolated antibody

Synonym(s):

Anti-CLABP, Anti-CLIPINA (Coronin-like protein A), Anti-CORO1A, Anti-HCORO1, Anti-TACO (Tryptophane aspartate-containing goat protein ), Anti-p57

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

antigen ~57 kDa

species reactivity

rat, mouse, human

packaging

antibody small pack of 25 μL

concentration

~1.5 mg/mL

technique(s)

western blot: 1.5-3.0 μg/mL using Jurkat cell lysates, or lysates of mouse brain (S1 fraction) or rat spleen (S1 fraction)

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... CORO1A(11151)

General description

Coronin 1C (CORO1C) is encoded by the gene mapped to human chromosome 12q24.1. The encoded protein belongs to coronin-like family of proteins. CORO1C consists of 474 amino acids and is ubiquitously expressed in human tissues. CORO1C contains conserved five WD repeats in its N-terminal end and a coiled-coil motif in its C-terminal end.The protein is mainly present at the sites of active actin dynamics including lamellipodia and membrane ruffles.

Specificity

Anti-Coronin-1A (C-terminal) specifically recognizes human, mouse, and rat coronin-1A.

Biochem/physiol Actions

Coronin 1C (CORO1C) plays a vital role in cytokinesis, motility and signal transduction. It is also implicated in regulation of various cellular processes such as cell proliferation, migration, invadopodia formation and invasion in glioblastoma cells. CORO1C acts as a predictive biomarker for hepatocellular carcinoma (HCC) invasive behavior. The encoded protein negatively regulates cell-matrix adhesion and intestinal epithelial cells (IECs) migration via FAK-mediated signaling.

Physical form

Solution in 0.01 M phos­phate buffered saline, pH 7.4, containing 15 mM sodium azide.

Storage and Stability

Store at –20 °C. For continuous use, the product may be stored at 2–8 °C for up to one month. For extended storage, freeze in working aliquots at –20 °C. Repeated freezing and thawing, or storage in “frost-free” freezers, is not recommended. If slight turbidity occurs upon prolonged storage, clarify the solution by centrifugation before use. Working dilutions should be discarded if not used within 12 hours.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class

10 - Combustible liquids

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Coronin-1C is a novel biomarker for hepatocellular carcinoma invasive progression identified by proteomics analysis and clinical validation.
Wu L
Journal of Experimental & Clinical Cancer Research (2010)
Coronin 1C negatively regulates cell-matrix adhesion and motility of intestinal epithelial cells.
Samarin SN
Biochemical and Biophysical Research Communications, 391, 394-400 (2010)
Isolation and chromosomal assignment of a novel human gene, CORO1C, homologous to coronin-like actin-binding proteins.
Iizaka M
Cytogenetics and Cell Genetics, 88, 221-224 (2000)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service