Skip to Content
MilliporeSigma
All Photos(3)

Key Documents

SAB4200076

Sigma-Aldrich

Anti-Nedd9 (C-terminal) antibody produced in rabbit

enhanced validation

affinity isolated antibody

Synonym(s):

Anti-Cas-L, Anti-HEF1

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

rabbit

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

antigen ~115 kDa

species reactivity

human

packaging

antibody small pack of 25 μL

enhanced validation

recombinant expression
Learn more about Antibody Enhanced Validation

technique(s)

indirect immunofluorescence: 1-2 μg/mL using human HS68 cells
western blot: 1.5-3.0 μg/mL using extracts of HEK-293T cells overexpressing human Nedd9

UniProt accession no.

shipped in

dry ice

Storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... NEDD9(4739)
mouse ... Nedd9(18003)
rat ... Nedd9(291044)

General description

Neural precursor cell expressed, developmentally down-regulated 9 (Nedd9), also known as Cas-L and human enhancer of filamentation 1 (HEF1) is an adhesion adapter protein, localized to sites of focal adhesions. It belongs to a larger family of Cas docking adapter proteins including p130Cas and Efs and is expressed preferentially in lymphocytes and epithelial cells. The gene NEDD9 is mapped to human chromosome 6p24.2.

Specificity

Anti-Nedd9 (C-terminal) specifically recognizes human Nedd9.

Application

Anti-Nedd9 (C-terminal) antibody produced in rabbit is suitable for immunoblotting and immunofluorescence.

Biochem/physiol Actions

Neural precursor cell expressed, developmentally down-regulated 9 (Nedd9) protein participates in integrin and growth factor signaling pathways and regulates growth, motility and apoptosis. It undergoes extensive phosphorylation, a process dependent both on cell adhesion and on the integrity of actin cytoskeleton. In response to integrin receptor binding to the extracellular matrix (ECM), Nedd9 binds to focal adhesion kinase (FAK) and is phosphorylated by FAK and Src kinases. In addition, Nedd9 is regulated at multiple levels in a complex and cell cycle-dependent manner. It is thought to play an important role in early neuronal differentiation and brain development. Nedd9 is also a key player in metastatic cancer. Upregulation of Nedd9 expression is observed in various solid tumors and is greatly implicated in the pathogenesis of BCR-Abl-dependent tumors. Overexpression of Nedd9 has been shown to be required for invasion by glioblastoma and to be strongly linked to the promotion of melanoma metastasis.

Physical form

Solution in 0.01 M posphate buffered saline, pH 7.4, containing 15 mM sodium azide.

Storage and Stability

Store at –20 °C. For continuous use, the product may be stored at 2–8 °C for up to one month. For extended storage, freeze in working aliquots at –20 °C. Repeated freezing and thawing, or storage in “frost-free” freezers, is not recommended. If slight turbidity occurs upon prolonged storage, clarify the solution by centrifugation before use. Working dilutions should be discarded if not used within 12 hours.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class

10 - Combustible liquids

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

High-resolution copy number analysis of clear cell endometrial carcinoma
O Hara AJ, et al.
Cancer Genetics, 240(, author=), 5-14 (2020)
NEDD9 stabilizes focal adhesions, increases binding to the extra-cellular matrix and differentially effects 2D versus 3D cell migration
Zhong J, et al.
Testing, 7(4), e35058-e35058 (2012)
The docking protein HEF1 is an apoptotic mediator at focal adhesion sites
Law SF, et al.
Molecular and Cellular Biology, 20(14), 5184-5195 (2000)
A new central scaffold for metastasis: parsing HEF1/Cas-L/NEDD9
O'Neill GM, et al.
Cancer Research, 67(19), 8975-8979 (2007)
Comparative oncogenomics identifies NEDD9 as a melanoma metastasis gene
Kim M, et al.
Cell, 125(7), 1269-1281 (2006)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service