Skip to Content
MilliporeSigma
All Photos(3)

Key Documents

SAB4200066

Sigma-Aldrich

Anti-EIF2C1/Ago1 (N-terminal) antibody produced in rabbit

~1.0 mg/mL, affinity isolated antibody, buffered aqueous solution

Synonym(s):

Anti-Argonaute-1, Anti-Eukaryotic translation initiation factor 2C, subunit 1, Anti-GERP95, Anti-Golgi Endoplasmic Reticulum protein 95 kDa

Sign Into View Organizational & Contract Pricing

Select a Size

200 μL
$562.00

$562.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
200 μL
$562.00

About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

$562.00


Check Cart for Availability

Request a Bulk Order

biological source

rabbit

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

antigen ~100 kDa

species reactivity

human

concentration

~1.0 mg/mL

technique(s)

immunoprecipitation (IP): 5-10 μg using lysates of HEK-293T cells over expressing hEIF2C1/Ago1
indirect immunofluorescence: 2-5 μg/mL using paraformaldehyde fixed HEK-293T cells over expressing hEIF2C1/Ago1
western blot: 0.5-1 μg/mL using lysates of HEK-293T cells over expressing hEIF2C1/Ago1

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

General description

In humans, the argonaute (Ago) subfamily consists of four isoforms hsAgo1−4. Ago proteins (~100 kDa molecular weight) contain a piwi-argonaute-zwille (PAZ) and PIWI domains. These proteins are evolutionarily conserved between species. EIF2C1/Ago1 is predominantly expressed in embryonic kidney and lung.
The eukaryotic initiation factor (EIF2C1), also known as argonaute 1, RNA-induced silencing complex (RISC) catalytic component, is encoded by the gene with 19 exons and 18 introns spanning ~ 50 kb on genomic DNA, mapped to human chromosome 1p34.3. The gene is widely expressed at low and medium levels. The encoded protein is a member of argonaute family of proteins.

Application

Anti-EIF2C1/Ago1 (N-terminal) antibody produced in rabbit has also been used in:
  • immunoblotting
  • immunoprecipitation
  • immunofluorescence

Anti-EIF2C1/Ago1 (N-terminal) antibody produced in rabbit has been used in chromatin immunoprecipitation (ChIP) and RNA-native ChIP (NChIP) experiments.

Biochem/physiol Actions

EIF2C1 is implicated in both transcriptional and post-transcriptional gene silencing. EIF2C1 levels are also increased in tumors that lack the Wilm′s tumor suppressor gene, WT1.
Eukaryotic initiation factor (EIF2C1)/ argonaute 1 plays a vital role in activation of transcriptional enhancers and also regulates alternative splicing in human cells. The encoded protein interacts with hypoxia-responsive microRNAs (HRMs) and stimulate angiogenic pathway.

Physical form

Solution in 0.01 M phos­phate buffered saline, pH 7.4, containing 15 mM sodium azide.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class

10 - Combustible liquids

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Hypoxia-responsive miRNAs target argonaute 1 to promote angiogenesis
Chen Z
The Journal of Clinical Investigation, 123, 1057-1067 (2013)
Argonaute proteins couple chromatin silencing to alternative splicing.
Ameyar-Zazoua M
Nature Structural and Molecular Biology, 19, 998-1004 (2012)
Argonaute proteins: potential biomarkers for human colon cancer
Li L, et al.
BMC Cancer, 10(1), 38-38 (2010)
Argonaute proteins at a glance
Ender C and Meister G
Journal of Cell Science, 123(11), 1819-1823 (2010)
Human eukaryotic initiation factor EIF2C1 gene: cDNA sequence, genomic organization, localization to chromosomal bands 1p34-p35, and expression.
Koesters R
Genomics, 61(2), 210-218 (1999)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service