Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

SAB4200063

Sigma-Aldrich

Anti-Snurportin 1 (C-terminal) antibody produced in rabbit

~1.0 mg/mL, affinity isolated antibody

Synonym(s):

Anti-RNA U transporter 1, Anti-RNUT1, Anti-SNUPN

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

rabbit

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

antigen ~41 kDa

packaging

antibody small pack of 25 μL

concentration

~1.0 mg/mL

technique(s)

immunoprecipitation (IP): 2.5-5 μg using lysates of HEK-293T cells over expressing human Snurportin 1
western blot: 1-2 μg/mL using lysates of HEK-293T cells over expressing human Snurportin 1

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... SNUPN(10073)

General description

Snurportin 1 contains an N-terminal importin β binding (IBB) motif, 2,2,7-trimethylguanosine (TMG) cap-binding domain, an ill-defined region and a C‐terminal m3Gcap-binding region. The SNUPN gene is mapped on the human chromosome at 15q24.2.

Specificity

Anti-Snurportin 1 recognizes human snurportin 1.

Application

Anti-Snurportin 1 (C-terminal) antibody produced in rabbit may be used in immunoblotting and immunoprecipitation.

Biochem/physiol Actions

Snurportin 1 mediates the nuclear import of 5′‐2,2,7‐terminal trimethylguanosine (m3G)-capped uridine-rich small nuclear RNPs (U snRNPs), an important step in the maturation of the spliceosome. This protein binds with importin β that mediates the interaction with the nuclear pore complex (NPC). It also interacts with m3G-cap but not with 5′‐terminal 7‐monomethylguanosine (m7G)-cap structures and chromosomal maintenance 1 (Crm1)/exportin 1. Snurportin 1 was found to accumulate in Cajal bodies after the import of U2 but not U1 snRNPs.

Physical form

Solution in 0.01 M phos­phate buffered saline, pH 7.4, containing 15 mM sodium azide.

Storage and Stability

Store at –20 °C. For continuous use, the product may be stored at 2–8 °C for up to one month. For extended storage, freeze in working aliquots at –20 °C. Repeated freezing and thawing, or storage in “frost-free” freezers, is not recommended. If slight turbidity occurs upon prolonged storage, clarify the solution by centrifugation before use. Working dilutions should be discarded if not used within 12 hours.

Disclaimer

Unless otherwise stated in our catalog, our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class

10 - Combustible liquids

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

J Huber et al.
The EMBO journal, 17(14), 4114-4126 (1998-07-22)
The nuclear import of the spliceosomal snRNPs U1, U2, U4 and U5, is dependent on the presence of a complex nuclear localization signal (NLS). The latter is composed of the 5'-2,2,7-terminal trimethylguanosine (m3G) cap structure of the U snRNA and
Jason K Ospina et al.
Molecular biology of the cell, 16(10), 4660-4671 (2005-07-21)
The initial steps of spliceosomal small nuclear ribonucleoprotein (snRNP) maturation take place in the cytoplasm. After formation of an Sm-core and a trimethylguanosine (TMG) cap, the RNPs are transported into the nucleus via the import adaptor snurportin1 (SPN) and the
Rosa Romano et al.
Clinical immunology (Orlando, Fla.), 176, 71-76 (2017-01-18)
We report the first patient with an interstitial deletion of chromosome 15q24.1-q24.3 associated with common variable immunodeficiency (CVID). The 18-year old female patient's clinical and immunological phenotype was compared with 8 additional previously published patients with chr15q24 deletions. A CGH

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service