Skip to Content
MilliporeSigma
All Photos(4)

Key Documents

SAB4200058

Sigma-Aldrich

Anti-Rab13 (C-terminal) antibody produced in rabbit

~1.0 mg/mL, affinity isolated antibody, buffered aqueous solution

Synonym(s):

Anti-GIG4, Anti-Growth-inhibiting gene 4 protein, Anti-RAS-associated protein RAB13

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

rabbit

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

antigen ~23 kDa

species reactivity

human

packaging

antibody small pack of 25 μL

concentration

~1.0 mg/mL

technique(s)

immunoprecipitation (IP): 5-10 μg using lysates of human NT2 cells
indirect immunofluorescence: 2-5 μg/mL using human A431 cells
western blot: 5-10 μg/mL using whole extracts of human NT2 cells

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... RAB13(5872)

General description

Rab (Ras-related in brain) 13 is encoded by the gene mapped to human chromosome 1q21.3. It is a member of Rab family of proteins. The encoded protein is ubiquitously expressed and is mainly localized at the trans-Golgi network, on recycling endosomes, on late endosomes, and at the plasma membrane.

Application

Anti-Rab13 (C-terminal) antibody produced in rabbit has been used in:
  • immunoprecipitation
  • immunofluorescence
  • western blotting

Biochem/physiol Actions

Rab (Ras-related in brain) 13 plays a vital role in both biosynthetic and endosomal recycling pathways. It is also involved in the regulation of membrane trafficking at the level of endosomes. Rab13 helps in the transport of cargo (integrins) to the plasma membrane, disruption of cell-cell adhesions. Additionally, it aids in cell migration by rearranging actin cytoskeleton attached to surface delivery of selective proteins. It also plays a crucial role in cancer progression. Rab13 is also involved in a putative secretory function in osteoclasts.

Physical form

Solution in 0.01 M phos­phate buffered saline, pH 7.4, containing 15 mM sodium azide.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class

10 - Combustible liquids

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Regulation of Cancer Cell Behavior by the Small GTPase Rab13.
Ioannou MS and McPherson PS
The Journal of Biological Chemistry, 291, 9929-9937 (2016)
Loss-of-function mutations in ATP6AP1 and ATP6AP2 in granular cell tumors
Pareja F, et al.
Nature Communications, 9(1), 3533-3533 (2018)
Predicting drug susceptibility of non-small cell lung cancers based on genetic lesions.
Sos ML
The Journal of Clinical Investigation, 119, 1727-1740 (2009)
Mirkka J Hirvonen et al.
The journal of histochemistry and cytochemistry : official journal of the Histochemistry Society, 60(7), 537-549 (2012-05-09)
Osteoclasts are bone-resorbing multinucleated cells that undergo drastic changes in their polarization due to heavy vesicular trafficking during the resorption cycle. These events require the precise orchestration of membrane traffic in order to maintain the unique characteristics of the different
Fresia Pareja et al.
Nature communications, 9(1), 3533-3533 (2018-09-01)
Granular cell tumors (GCTs) are rare tumors that can arise in multiple anatomical locations, and are characterized by abundant intracytoplasmic granules. The genetic drivers of GCTs are currently unknown. Here, we apply whole-exome sequencing and targeted sequencing analysis to reveal

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service